Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Alkbh7 PubMed Links |
Alkbh7 - alkB, alkylation repair homolog 7 (E. coli) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Clpp - caseinolytic mitochondrial matrix peptidase proteolytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017393.2 | 1105 | UTR 3 | NP_059089.1 |
Gm33509 - predicted gene, 33509 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Gtf2f1 - general transcription factor IIF, polypeptide 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Pspn - persephin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |