Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Chrnd PubMed Links |
Chrnd - cholinergic receptor, nicotinic, delta polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021600.3 | Intron | NP_067611.2 |
Chrng - cholinergic receptor, nicotinic, gamma polypeptide | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Prss56 - protease, serine 56 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |