Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGAGCCCTTCTCAGATGTCCCC[A/G]AGTAAGGAGACTGGCAAAATAAGGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Galnt10 PubMed Links |
Galnt10 - UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_134189.2 | Intron | NP_598950.2 |