Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCCAGACCTTTTCTGCTGGGTC[A/T]TTCTTCTCAACATCCAAAAATGCCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pitpnm3 PubMed Links |
Pitpnm3 - PITPNM family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024927.2 | Intron | NP_001020098.1 | ||||
NM_001081641.1 | Intron | NP_001075110.1 |