Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCAACAGCGTCTCCCCCGTACC[C/T]GAAGCTGTGGAGGTGGGTGCTATAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Htr5a PubMed Links |
Htr5a - 5-hydroxytryptamine (serotonin) receptor 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008314.2 | 1231 | Silent Mutation | NP_032340.2 |