Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM11296
          Assay Name: hsa-miR-487b-3p
          miRBase Accession Number: MI0003530
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101046455 - 101046538 [+] on Build GRCh38
          Mature miRNA Sequence: AAUCGUACAGGGUCAUCCACUU
          Species: Human, Mouse, Rat, Bovine, Callithrix jacchus, Dasypus novemcinctus, Dog, Goat, Guinea pig, Horse, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Sheep
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM11296
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000018, mir-154

          Mature miRNA Sequence:

          AAUCGUACAGGGUCAUCCACUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-487b-3p MIMAT0003180
          Mouse mmu-miR-487b-3p MIMAT0003184
          Rat rno-miR-487b-3p MIMAT0003200
          Bovine bta-miR-487b MIMAT0003847
          Callithrix jacchus cja-miR-487b MIMAT0039762
          Dasypus novemcinctus dno-miR-487b-3p MIMAT0047917
          View More Rows
          Dog cfa-miR-487b MIMAT0006723
          Goat chi-miR-487b-3p MIMAT0036249
          Guinea pig cpo-miR-487b-3p MIMAT0047282
          Horse eca-miR-487b MIMAT0013162
          Pan troglodytes ptr-miR-487b MIMAT0008162
          Pongo pygmaeus ppy-miR-487b MIMAT0015906
          Pteropus alecto pal-miR-487b-3p MIMAT0040020
          Rabbit ocu-miR-487b-3p MIMAT0048474
          Rhesus monkey mml-miR-487b-3p MIMAT0006347
          Sheep oar-miR-487b-3p MIMAT0019295

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-487b
          Stem-loop Accession # MI0003530
          Stem-loop Sequence
          UUGGUACUUGGAGAGUGGUUAUCCCUGUCCUGUUCGUUUUGCUCAUGUCGAAUCGUACAGGGUCAUCCACUUUUUCAGUAUCAA
          Chromosome Location Chr. 14 - 101046455 - 101046538 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003180
          miRBase ID: hsa-miR-487b-3p
          Mature miRNA Sequence: AAUCGUACAGGGUCAUCCACUU
          Chromosome Location: Chr. 14 - 101046455 - 101046538 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-487b
          Stem-loop Accession # MI0003534
          Stem-loop Sequence
          UGGUACUUGGAGAGUGGUUAUCCCUGUCCUCUUCGCUUCACUCAUGCCGAAUCGUACAGGGUCAUCCACUUUUUCAGUAUCA
          Chromosome Location Chr. 12 - 109727333 - 109727414 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003184
          miRBase ID: mmu-miR-487b-3p
          Mature miRNA Sequence: AAUCGUACAGGGUCAUCCACUU
          Chromosome Location: Chr. 12 - 109727333 - 109727414 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-487b
          Stem-loop Accession # MI0003547
          Stem-loop Sequence
          UGGUACUUGGAGAGUGGUUAUCCCUGUCCUCUUCGCUUCACUUAUGCCGAAUCGUACAGGGUCAUCCACUUUUUCAGUAUCA
          Chromosome Location Chr. 6 - 133877124 - 133877205 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0003200
          miRBase ID: rno-miR-487b-3p
          Mature miRNA Sequence: AAUCGUACAGGGUCAUCCACUU
          Chromosome Location: Chr. 6 - 133877124 - 133877205 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-487b
          Stem-loop Accession # MI0005060
          Stem-loop Sequence
          UUGGUACUUGGAGAGUGGUUAUCCCUGUCCUGUUCGUUUUACUCAUGUCGAAUCGUACAGGGUCAUCCACUUUUUCAGUAUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003847
          miRBase ID: bta-miR-487b
          Mature miRNA Sequence: AAUCGUACAGGGUCAUCCACUU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-487b
          Stem-loop Accession # MI0032312
          Stem-loop Sequence
          GGCUUUCCUUUCCGUGCUAACCUUUGGUACUUGGAGAGUGGUUAUCCCUGUCCUGUUCGUUUUGCUCAUGUCGAAUCGUACAGGGUCAUCCACUUUUUCAGUAUCAAGAGCGCAGACUCUGGAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039762
          miRBase ID: cja-miR-487b
          Mature miRNA Sequence: AAUCGUACAGGGUCAUCCACUU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-487b
          Stem-loop Accession # MI0039124
          Stem-loop Sequence
          GUGGUUAUCCCUGUCCCGUUCGUUUUACUUGUGUCGAAUCGUACAGGGUCAUCCACUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047917
          miRBase ID: dno-miR-487b-3p
          Mature miRNA Sequence: AAUCGUACAGGGUCAUCCACUU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-487b
          Stem-loop Accession # MI0008144
          Stem-loop Sequence
          GUGGUUAUCCCUGUCCUGUUCGUUUUACUCAUGUCGAAUCGUACAGGGUCAUCCACUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006723
          miRBase ID: cfa-miR-487b
          Mature miRNA Sequence: AAUCGUACAGGGUCAUCCACUU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-487b
          Stem-loop Accession # MI0030804
          Stem-loop Sequence
          UUUCUUUCCCGUGCUAACCUUUGGUACUUGGAGAGUGGUUAUCCCUGUCCUGUUCGUUUUACUCAUGUCGAAUCGUACAGGGUCAUCCACUUUUUCAGUAUCAAGAGCGCAGACCGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036249
          miRBase ID: chi-miR-487b-3p
          Mature miRNA Sequence: AAUCGUACAGGGUCAUCCACUU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-487b
          Stem-loop Accession # MI0038790
          Stem-loop Sequence
          GUGGUUAUCCCUGUCCUGUUCGUUUCACUCACGUCGAAUCGUACAGGGUCAUCCACUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047282
          miRBase ID: cpo-miR-487b-3p
          Mature miRNA Sequence: AAUCGUACAGGGUCAUCCACUU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-487b
          Stem-loop Accession # MI0012905
          Stem-loop Sequence
          UUGGUACUUGGAGAGUGGUUAUCCCUGUCCUGUUCGUUUUAAUUACGUCGAAUCGUACAGGGUCAUCCACUUUUUCAGUAUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013162
          miRBase ID: eca-miR-487b
          Mature miRNA Sequence: AAUCGUACAGGGUCAUCCACUU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-487b
          Stem-loop Accession # MI0008684
          Stem-loop Sequence
          UGGUACUUGGAGAGUGGUUAUCCCUGUCCUGUUCGUUUUGCUCAUGUCGAAUCGUACAGGGUCAUCCACUUUUUCAGUAUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008162
          miRBase ID: ptr-miR-487b
          Mature miRNA Sequence: AAUCGUACAGGGUCAUCCACUU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-487b
          Stem-loop Accession # MI0014960
          Stem-loop Sequence
          UUGGUACUUGGAGAGUGGUUAUCCCUGUCCUGUUCGUUUUGCUCAUGUCGAAUCGUACAGGGUCAUCCACUUUUUCAGUAUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015906
          miRBase ID: ppy-miR-487b
          Mature miRNA Sequence: AAUCGUACAGGGUCAUCCACUU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-487b
          Stem-loop Accession # MI0032451
          Stem-loop Sequence
          GUGGUUAUCCCUGUCCUGUUCGUUUUACUCAUGUCGAAUCGUACAGGGUCAUCCACUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040020
          miRBase ID: pal-miR-487b-3p
          Mature miRNA Sequence: AAUCGUACAGGGUCAUCCACUU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-487b
          Stem-loop Accession # MI0039419
          Stem-loop Sequence
          GUGGUUAUCCCUGUCCUGUUCGUUUUAUUCAUGUCGAAUCGUACAGGGUCAUCCACUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048474
          miRBase ID: ocu-miR-487b-3p
          Mature miRNA Sequence: AAUCGUACAGGGUCAUCCACUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-487b
          Stem-loop Accession # MI0007761
          Stem-loop Sequence
          UUGGUACUUGGAGAGUGGUUAUCCCUGUCCUGUUCGUUUUGCUCGUGUCGAAUCGUACAGGGUCAUCCACUUUUUCAGUAUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006347
          miRBase ID: mml-miR-487b-3p
          Mature miRNA Sequence: AAUCGUACAGGGUCAUCCACUU
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-487b
          Stem-loop Accession # MI0016941
          Stem-loop Sequence
          UGCUAACCUUUGGUACUUGGAGAGUGGUUAUCCCUGUCCUGUUCGUUUUACUCAUGUCGAAUCGUACAGGGUCAUCCACUUUUUCAGUAUCAAGAGCGCAGACCGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019295
          miRBase ID: oar-miR-487b-3p
          Mature miRNA Sequence: AAUCGUACAGGGUCAUCCACUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307703_pri
          TaqMan™ Pri-miRNA Assay : Hs03303552_pri
          TaqMan™ Pri-miRNA Assay : Rn03464898_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11296
          TaqMan™ MicroRNA Assay : 001285
          mirVana® miRNA inhibitor : MH11296
          mirVana® miRNA mimic : MC11296
          TaqMan™ Advanced miRNA Assay : mmu481184_mir
          TaqMan™ Advanced miRNA Assay : 477835_mir
          TaqMan™ Advanced miRNA Assay : rno481184_mir


          miRBase Alias:

          Human: hsa-miR-487b (v19)
          Mouse: mmu-miR-487b (v17)
          Rat: rno-miR-487b (v18)
          Citrobacter farmeri: cfa-miR-487 (v12)
          Macaca mulatta: mml-miR-487b (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline