Search
Search

| Assay Name: | rno-miR-423-5p |
|---|---|
| miRBase Accession Number: | MI0006145 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGAGGGGCAGAGAGCGAGACUUUU |
| Species: | Rat, Eptesicus fuscus |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000329, mir-423 |
Mature miRNA Sequence: | UGAGGGGCAGAGAGCGAGACUUUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-423-5p | MIMAT0017305 |
| Eptesicus fuscus | efu-miR-423 | MIMAT0035024 |