Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM11174
          Assay Name: hsa-let-7a-2-3p
          miRBase Accession Number: MI0000061
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 122146522 - 122146593 [-] on Build GRCh38
          Mature miRNA Sequence: CUGUACAGCCUCCUAGCUUUCC
          Species: Human, Rat, Alligator mississippiensis, Anolis carolinensis, Chrysemys picta, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Monodelphis domestica, Ophiophagus hannah, Pteropus alecto, Rhesus monkey, Salmo salar
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM11174
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000002, let-7

          Mature miRNA Sequence:

          CUGUACAGCCUCCUAGCUUUCC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-let-7a-2-3p MIMAT0010195
          Rat rno-let-7a-2-3p MIMAT0017086
          Alligator mississippiensis ami-let-7c-2-3p MIMAT0038007
          Anolis carolinensis aca-let-7c-2-3p MIMAT0021698
          Chrysemys picta cpi-let-7c-2-3p MIMAT0037601
          Cricetulus griseus cgr-let-7a-2 MIMAT0041130
          View More Rows
          Dasypus novemcinctus dno-let-7c-2-3p MIMAT0047476
          Guinea pig cpo-let-7a-2-3p MIMAT0046819
          Monodelphis domestica mdo-let-7a-3p MIMAT0026686
          Ophiophagus hannah oha-let-7c-1-3p MIMAT0036631
          Pteropus alecto pal-let-7a-2-3p MIMAT0040148
          Rhesus monkey mml-let-7a-2-3p MIMAT0031041
          Salmo salar ssa-let-7a-3p MIMAT0032689

          Stem-loop Details



          Human

          Stem-loop ID hsa-let-7a-2
          Stem-loop Accession # MI0000061
          Stem-loop Sequence
          AGGUUGAGGUAGUAGGUUGUAUAGUUUAGAAUUACAUCAAGGGAGAUAACUGUACAGCCUCCUAGCUUUCCU
          Chromosome Location Chr. 11 - 122146522 - 122146593 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0010195
          miRBase ID: hsa-let-7a-2-3p
          Mature miRNA Sequence: CUGUACAGCCUCCUAGCUUUCC
          Chromosome Location: Chr. 11 - 122146522 - 122146593 [-] on Build GRCh38

          Rat

          Stem-loop ID rno-let-7a-2
          Stem-loop Accession # MI0000828
          Stem-loop Sequence
          CGGCAUGCUCCCAGGCUGAGGUAGUAGGUUGUAUAGUUUAGAGUUACAACAAGGGAGAUAACUGUACAGCCUCCUAGCUUUCCUUGGGACUUGCAC
          Chromosome Location Chr. 8 - 45753264 - 45753359 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017086
          miRBase ID: rno-let-7a-2-3p
          Mature miRNA Sequence: CUGUACAGCCUCCUAGCUUUCC
          Chromosome Location: Chr. 8 - 45753264 - 45753359 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-let-7c-2
          Stem-loop Accession # MI0029645
          Stem-loop Sequence
          GGGUGGGGGCCGGGGUGAGGUAGUAGGUUGUAUGGUUUAGAGUCACACCCCCGGAGAUAACUGUACAGCCUCCUAGCUUUCCCCGCGCCCCCCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038007
          miRBase ID: ami-let-7c-2-3p
          Mature miRNA Sequence: CUGUACAGCCUCCUAGCUUUCC
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-let-7c-2
          Stem-loop Accession # MI0018704
          Stem-loop Sequence
          CCGGCGUGCCUCGAGUUGAGGUAGUAGGUUGUAUGGUUUAGAAUAACACCAUCAGGAGAUAACUGUACAGCCUCCUAGCUUUCCCCGGGGCAACGCCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021698
          miRBase ID: aca-let-7c-2-3p
          Mature miRNA Sequence: CUGUACAGCCUCCUAGCUUUCC
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-let-7c-2
          Stem-loop Accession # MI0029377
          Stem-loop Sequence
          UGCGUGCCCCAGGCUGAGGUAGUAGGUUGUAUGGUUUAGAAUAACACCCGCAGGAGAUAACUGUACAGCCUCCUAGCUUUCCCUGGGGCGCUGCUUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037601
          miRBase ID: cpi-let-7c-2-3p
          Mature miRNA Sequence: CUGUACAGCCUCCUAGCUUUCC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-let-7a-2
          Stem-loop Accession # MI0033058
          Stem-loop Sequence
          AGGUUGAGGUAGUAGGUUGUAUAGUUUAGAGUUACAUCAAGGGAGAUAACUGUACAGCCUCCUAGCUUUCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041130
          miRBase ID: cgr-let-7a-2
          Mature miRNA Sequence: CUGUACAGCCUCCUAGCUUUCC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-let-7c-2
          Stem-loop Accession # MI0038887
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUAUGGUUUAGAAUUACAUCAAGGGAGAUAACUGUACAGCCUCCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047476
          miRBase ID: dno-let-7c-2-3p
          Mature miRNA Sequence: CUGUACAGCCUCCUAGCUUUCC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-let-7a-2
          Stem-loop Accession # MI0038541
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUAUAGUUUAGAAUUACAUCAAGGGAGAUAACUGUACAGCCUCCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046819
          miRBase ID: cpo-let-7a-2-3p
          Mature miRNA Sequence: CUGUACAGCCUCCUAGCUUUCC
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-let-7a-2
          Stem-loop Accession # MI0005330
          Stem-loop Sequence
          AGGUUGAGGUAGUAGGUUGUAUAGUUUAGAAUUACAUCAAGGGAGAUAACUGUACAGCCUCCUAGCUUUCCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026686
          miRBase ID: mdo-let-7a-3p
          Mature miRNA Sequence: CUGUACAGCCUCCUAGCUUUCC
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-let-7c-1
          Stem-loop Accession # MI0031302
          Stem-loop Sequence
          AAGCUGAGGUAGUAGGUUGUAUGGUUUAGAAUUACACCAAGGGAGAUAACUGUACAGCCUCCUAGCUUUCCUUGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036631
          miRBase ID: oha-let-7c-1-3p
          Mature miRNA Sequence: CUGUACAGCCUCCUAGCUUUCC
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-let-7a-2
          Stem-loop Accession # MI0032518
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUAUAGUUUAGAAUUGCAUCAAGGGAGAUAACUGUACAGCCUCCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040148
          miRBase ID: pal-let-7a-2-3p
          Mature miRNA Sequence: CUGUACAGCCUCCUAGCUUUCC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-let-7a-2
          Stem-loop Accession # MI0007571
          Stem-loop Sequence
          AGGCUGAGGUAGUAGGUUGUAUAGUUUAGAAUUACAUCAAGGGAGAUAACUGUACAGCCUCCUAGCUUUCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031041
          miRBase ID: mml-let-7a-2-3p
          Mature miRNA Sequence: CUGUACAGCCUCCUAGCUUUCC
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-let-7a-1
          Stem-loop Accession # MI0026740
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUAUAGUUCAGAGUGACAUCACAGGAGAUAACUGUACAGCCUCCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032689
          miRBase ID: ssa-let-7a-3p
          Mature miRNA Sequence: CUGUACAGCCUCCUAGCUUUCC
          Chromosome Location: NA

          Stem-loop ID ssa-let-7a-2
          Stem-loop Accession # MI0026742
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUAUAGUUGAGAAUUACACCCCGGGAGAUAACUGUACAGCCUCCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032689
          miRBase ID: ssa-let-7a-3p
          Mature miRNA Sequence: CUGUACAGCCUCCUAGCUUUCC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465205_pri
          TaqMan™ Pri-miRNA Assay : Hs03302539_pri
          Ambion® Pre-miR™ miRNA Precursor : PM11174
          TaqMan™ MicroRNA Assay : 121214_mat
          mirVana® miRNA inhibitor : MH11174
          mirVana® miRNA mimic : MC11174
          TaqMan™ Advanced miRNA Assay : 479269_mir


          miRBase Alias:

          Human: hsa-let-7a-2* (v17)
          Rat: rno-let-7a-2* (v18)
          Aeromonas caviae Ae398: aca-let-7c-2* (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b6cf8ccb7-mmm2n:80/100.66.78.224:80.
          git-commit: 76b2fce788d0185f5e22c122d85c407206860d42
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.0-2026-offline-prod