Search
Search

| Assay Name: | hsa-miR-330-5p |
|---|---|
| miRBase Accession Number: | MI0000803 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCUCUGGGCCUGUGUCUUAGGC |
| Species: | Human, Mouse, Rat, Dog, Horse, Pongo pygmaeus, Rhesus monkey |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000200, mir-330 |
Mature miRNA Sequence: | UCUCUGGGCCUGUGUCUUAGGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-330-5p | MIMAT0004693 |
| Mouse | mmu-miR-330-5p | MIMAT0004642 |
| Rat | rno-miR-330-5p | MIMAT0004641 |
| Dog | cfa-miR-330 | MIMAT0009893 |
| Horse | eca-miR-330 | MIMAT0013012 |
| Pongo pygmaeus | ppy-miR-330-5p | MIMAT0015833 |
| View More Rows | ||
| Rhesus monkey | mml-miR-330-5p | MIMAT0006270 |