Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM11778
          Assay Name: hsa-let-7d-5p
          miRBase Accession Number: MI0000065
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 94178834 - 94178920 [+] on Build GRCh38
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Species: Human, Mouse, Rat, Bovine, Callithrix jacchus, Chrysemys picta, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Horse, Ophiophagus hannah, Ornithorhynchus anatinus, Pan troglodytes, Pig, Pongo pygmaeus, Python bivittatus, Rabbit, Rhesus monkey, Taeniopygia guttata
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM11778
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000002, let-7

          Mature miRNA Sequence:

          AGAGGUAGUAGGUUGCAUAGUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-let-7d-5p MIMAT0000065
          Mouse mmu-let-7d-5p MIMAT0000383
          Rat rno-let-7d-5p MIMAT0000562
          Bovine bta-let-7d MIMAT0003810
          Callithrix jacchus cja-let-7d MIMAT0049327
          Chrysemys picta cpi-let-7d-5p MIMAT0037602
          View More Rows
          Columba livia cli-let-7d-5p MIMAT0038381
          Cricetulus griseus cgr-let-7d-5p MIMAT0023719
          Dasypus novemcinctus dno-let-7d-5p MIMAT0047480
          Goat chi-let-7d-5p MIMAT0035886
          Guinea pig cpo-let-7d-5p MIMAT0046824
          Horse eca-let-7d MIMAT0013110
          Ophiophagus hannah oha-let-7d-5p MIMAT0036636
          Ornithorhynchus anatinus oan-let-7d-5p MIMAT0007234
          Pan troglodytes ptr-let-7d MIMAT0007939
          Pig ssc-let-7d-5p MIMAT0025356
          Pongo pygmaeus ppy-let-7d MIMAT0015724
          Python bivittatus pbv-let-7d-5p MIMAT0038801
          Rabbit ocu-let-7d-5p MIMAT0048082
          Rhesus monkey mml-let-7d MIMAT0006154
          Taeniopygia guttata tgu-let-7d-5p MIMAT0014536

          Stem-loop Details



          Human

          Stem-loop ID hsa-let-7d
          Stem-loop Accession # MI0000065
          Stem-loop Sequence
          CCUAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGGGAUUUUGCCCACAAGGAGGUAACUAUACGACCUGCUGCCUUUCUUAGG
          Chromosome Location Chr. 9 - 94178834 - 94178920 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000065
          miRBase ID: hsa-let-7d-5p
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: Chr. 9 - 94178834 - 94178920 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-let-7d
          Stem-loop Accession # MI0000405
          Stem-loop Sequence
          AAUGGGUUCCUAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGAGAUUUUGCCCACAAGGAGUUAACUAUACGACCUGCUGCCUUUCUUAGGGCCUUAUU
          Chromosome Location Chr. 13 - 48536012 - 48536114 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000383
          miRBase ID: mmu-let-7d-5p
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: Chr. 13 - 48536012 - 48536114 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-let-7d
          Stem-loop Accession # MI0000601
          Stem-loop Sequence
          UGGGCUCCUAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGAGAUUUUGCCCACAAGGAGUUAACUAUACGACCUGCUGCCUUUCUUAGGGCCUU
          Chromosome Location Chr. 17 - 16419980 - 16420077 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000562
          miRBase ID: rno-let-7d-5p
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: Chr. 17 - 16419980 - 16420077 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-let-7d
          Stem-loop Accession # MI0005026
          Stem-loop Sequence
          CCUAGGAAGAGGUAGUAGGUUGCAUAGUUUUCGGGCAGGGAUUUUGCCCACAAGGAGGUAACUAUACGACCUGCUGCCUUUCUUAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003810
          miRBase ID: bta-let-7d
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-let-7d
          Stem-loop Accession # MI0040063
          Stem-loop Sequence
          AGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGGGAUUUUGCCCACAAGGAGCUAACUAUACGACCUGCUGCCUUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049327
          miRBase ID: cja-let-7d
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-let-7d
          Stem-loop Accession # MI0029378
          Stem-loop Sequence
          UAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGAGAUUUUGCUCACAAGGAGUUAACUAUACAACCUGCUGCCUUUCUUAGGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037602
          miRBase ID: cpi-let-7d-5p
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-let-7d
          Stem-loop Accession # MI0029888
          Stem-loop Sequence
          UAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGGGAUUUUGCUCACAAGGAGGUAACUAUACAACCUGCUGCCUUUCUUAGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038381
          miRBase ID: cli-let-7d-5p
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-let-7d
          Stem-loop Accession # MI0020371
          Stem-loop Sequence
          GCUCCUAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGGGAUUUUGCCCACAAGGAGGUAACUAUACGACCUGCUGCCUUUCUUAGGGCCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023719
          miRBase ID: cgr-let-7d-5p
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-let-7d
          Stem-loop Accession # MI0038890
          Stem-loop Sequence
          AGAGGUAGUAGGUUGCAUAGUUUUGGGGCAGGGAUUUUGCCCACAAGGAGGUAACUAUACGACCUGCUGCCUUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047480
          miRBase ID: dno-let-7d-5p
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-let-7d
          Stem-loop Accession # MI0030582
          Stem-loop Sequence
          AAUGGGUUCCUAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGGGAUUUUGCCCACAAGGAGGUAACUAUACGACCUGCUGCCUUUCUUAGGGCCUUAUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035886
          miRBase ID: chi-let-7d-5p
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-let-7d
          Stem-loop Accession # MI0038545
          Stem-loop Sequence
          AGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGGGAUUUUGCCCACAAGGAGGUAACUAUACGACCUGCUGCCUUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046824
          miRBase ID: cpo-let-7d-5p
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-let-7d
          Stem-loop Accession # MI0012859
          Stem-loop Sequence
          CCUAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGGGAUUUUGCCCACAAGGAGAUAACUAUACGACCUGCUGCCUUUCUUAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013110
          miRBase ID: eca-let-7d
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-let-7d
          Stem-loop Accession # MI0031306
          Stem-loop Sequence
          GGGUUCCUAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGUGGUUUUGCCUAUACGGAGUUAACUAUACAACCUGCUGCCUUUCUUAGGGCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036636
          miRBase ID: oha-let-7d-5p
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-let-7d
          Stem-loop Accession # MI0006934
          Stem-loop Sequence
          GAAGAAACUAUGGGCUCCUAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGUGAUUUUGCUCAUAAGGUGUUAACUAUACAACCUGCUGCCUUUCUUAGGGCUUUAUUAUUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007234
          miRBase ID: oan-let-7d-5p
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-let-7d
          Stem-loop Accession # MI0008402
          Stem-loop Sequence
          CUAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGGGAUUUUGCCCACAAGGAGGUAACUAUACGACCUGCUGCCUUUCUUAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007939
          miRBase ID: ptr-let-7d
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-let-7d
          Stem-loop Accession # MI0022120
          Stem-loop Sequence
          GUUCCUAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGAGAUUUUGCCCACAAGGAGGUAACUAUACGACCUGCUGCCUUUCUUAGGGCCUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025356
          miRBase ID: ssc-let-7d-5p
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-let-7d
          Stem-loop Accession # MI0014784
          Stem-loop Sequence
          CCUAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGGGAUUUUGCCCACAAGGAGGUAACUAUACGACCUGCUGCCUUUCUUAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015724
          miRBase ID: ppy-let-7d
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-let-7d
          Stem-loop Accession # MI0030137
          Stem-loop Sequence
          GGGUUCCUAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGUGGUUUUGCCUAUAAGGAGUUAACUAUACAACCUGCUGCCUUUCUUAGGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038801
          miRBase ID: pbv-let-7d-5p
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-let-7d
          Stem-loop Accession # MI0039210
          Stem-loop Sequence
          AGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGGGAUUUUGCCCACAAGGAGGUAACUAUACGACCUGCUGCCUUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048082
          miRBase ID: ocu-let-7d-5p
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-let-7d
          Stem-loop Accession # MI0007575
          Stem-loop Sequence
          CCUAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGGGAUUUUGCCCACAAGGAGGUAACUAUACGACCUGCUGCCUUUCUUAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006154
          miRBase ID: mml-let-7d
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-let-7d
          Stem-loop Accession # MI0013750
          Stem-loop Sequence
          GCUCCUAGGAAGAGGUAGUAGGUUGCAUAGUUUUAGGGCAGGGAUUUUGCUCAGAAGGAGGUAACUAUACAACCUGCUGCCUUUCUUAGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014536
          miRBase ID: tgu-let-7d-5p
          Mature miRNA Sequence: AGAGGUAGUAGGUUGCAUAGUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465667_pri
          TaqMan™ Pri-miRNA Assay : Hs03302562_pri
          TaqMan™ Pri-miRNA Assay : Mm03306666_pri
          Ambion® Pre-miR™ miRNA Precursor : PM11778
          TaqMan™ MicroRNA Assay : 002283
          mirVana® miRNA inhibitor : MH11778
          mirVana® miRNA mimic : MC11778
          TaqMan™ Advanced miRNA Assay : rno478439_mir
          TaqMan™ Advanced miRNA Assay : mmu478439_mir
          TaqMan™ Advanced miRNA Assay : 478439_mir


          miRBase Alias:

          Human: hsa-let-7d (v17)
          Mouse: mmu-let-7d (v17)
          Rat: rno-let-7d (v18)
          Ochrobactrum anthropi: oan-let-7d (v18)
          Trabulsiella guamensis ATCC 49490: tgu-let-7d (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0