Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM12246
          Assay Name: hsa-miR-17-3p
          miRBase Accession Number: MI0000071
          miRBase Version: v22.1
          Chromosome Location: Chr.13: 91350605 - 91350688 [+] on Build GRCh38
          Mature miRNA Sequence: ACUGCAGUGAAGGCACUUGUAG
          Species: Human, Anolis carolinensis, Dasypus novemcinctus, Dog, Guinea pig, Ophiophagus hannah, Pig, Rabbit, Taeniopygia guttata
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM12246
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000001, mir-17

          Mature miRNA Sequence:

          ACUGCAGUGAAGGCACUUGUAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-17-3p MIMAT0000071
          Anolis carolinensis aca-miR-17-3p MIMAT0021804
          Dasypus novemcinctus dno-miR-17-3p MIMAT0047510
          Dog cfa-miR-17 MIMAT0006649
          Guinea pig cpo-miR-17-3p MIMAT0046858
          Ophiophagus hannah oha-miR-17-3p MIMAT0036752
          View More Rows
          Pig ssc-miR-17-3p MIMAT0015268
          Rabbit ocu-miR-17-3p MIMAT0048110
          Taeniopygia guttata tgu-miR-17a-3p MIMAT0014564

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-17
          Stem-loop Accession # MI0000071
          Stem-loop Sequence
          GUCAGAAUAAUGUCAAAGUGCUUACAGUGCAGGUAGUGAUAUGUGCAUCUACUGCAGUGAAGGCACUUGUAGCAUUAUGGUGAC
          Chromosome Location Chr. 13 - 91350605 - 91350688 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000071
          miRBase ID: hsa-miR-17-3p
          Mature miRNA Sequence: ACUGCAGUGAAGGCACUUGUAG
          Chromosome Location: Chr. 13 - 91350605 - 91350688 [+] on Build GRCh38

          Anolis carolinensis

          Stem-loop ID aca-mir-17
          Stem-loop Accession # MI0018772
          Stem-loop Sequence
          AUGUCAAAGUGCUUACAGUGCAGGUAGUGAUAAGUAGAACCUACUGCAGUGAAGGCACUUGUAGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021804
          miRBase ID: aca-miR-17-3p
          Mature miRNA Sequence: ACUGCAGUGAAGGCACUUGUAG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-17
          Stem-loop Accession # MI0038909
          Stem-loop Sequence
          CAAAGUGCUUACAGUGCAGGUAGUGAUAUGUGCAUCUACUGCAGUGAAGGCACUUGUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047510
          miRBase ID: dno-miR-17-3p
          Mature miRNA Sequence: ACUGCAGUGAAGGCACUUGUAG
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-17
          Stem-loop Accession # MI0008050
          Stem-loop Sequence
          CAAAGUGCUUACAGUGCAGGUAGUGAUAUGUGCAUCUACUGCAGUGAAGGCACUUGUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006649
          miRBase ID: cfa-miR-17
          Mature miRNA Sequence: ACUGCAGUGAAGGCACUUGUAG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-17
          Stem-loop Accession # MI0038565
          Stem-loop Sequence
          CAAAGUGCUUACAGUGCAGGUAGUGAUAUGUGCAUCUACUGCAGUGAAGGCACUUGUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046858
          miRBase ID: cpo-miR-17-3p
          Mature miRNA Sequence: ACUGCAGUGAAGGCACUUGUAG
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-17
          Stem-loop Accession # MI0031372
          Stem-loop Sequence
          UCAUGUUAGAAUAAUGUCAAAGUGCUUACAGUGCAGGUAGUGAUUUAUAGAACCUACUGCAGUGAAGGCACUUGUAGCAUUAUGCUGGCAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036752
          miRBase ID: oha-miR-17-3p
          Mature miRNA Sequence: ACUGCAGUGAAGGCACUUGUAG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-17
          Stem-loop Accession # MI0008214
          Stem-loop Sequence
          GAAUAAUGUCAAAGUGCUUACAGUGCAGGUAGUGAUAAUGUGCAUCUACUGCAGUGAAGGCACUUGUAGCAUUAUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015268
          miRBase ID: ssc-miR-17-3p
          Mature miRNA Sequence: ACUGCAGUGAAGGCACUUGUAG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-17
          Stem-loop Accession # MI0039225
          Stem-loop Sequence
          CAAAGUGCUUACAGUGCAGGUAGUGAUAUGUGCAUCUACUGCAGUGAAGGCACUUGUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048110
          miRBase ID: ocu-miR-17-3p
          Mature miRNA Sequence: ACUGCAGUGAAGGCACUUGUAG
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-17a
          Stem-loop Accession # MI0013784
          Stem-loop Sequence
          GUCAAAGUGCUUACAGUGCAGGUAGUGAUAUAUAGAACCUACUGCAGUGAAGGCACUUGUAGCAUUAUGUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014564
          miRBase ID: tgu-miR-17a-3p
          Mature miRNA Sequence: ACUGCAGUGAAGGCACUUGUAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03295901_pri
          Ambion® Pre-miR™ miRNA Precursor : PM12246
          TaqMan™ MicroRNA Assay : 002421
          mirVana® miRNA inhibitor : MH12246
          mirVana® miRNA mimic : MC12246
          TaqMan™ Advanced miRNA Assay : 477932_mir


          miRBase Alias:

          Human: hsa-miR-17* (v17)
          Aeromonas caviae Ae398: aca-miR-17* (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch