Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM12283
          Assay Name: mmu-miR-18a-3p
          miRBase Accession Number: MI0000567
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 115043851 - 115043946 [+] on Build GRCm38
          Mature miRNA Sequence: ACUGCCCUAAGUGCUCCUUCUG
          Species: Mouse, Alligator mississippiensis, Anolis carolinensis, Chicken, Chrysemys picta, Ophiophagus hannah, Python bivittatus
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM12283
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000001, mir-17

          Mature miRNA Sequence:

          ACUGCCCUAAGUGCUCCUUCUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-18a-3p MIMAT0004626
          Alligator mississippiensis ami-miR-18-3p MIMAT0038050
          Anolis carolinensis aca-miR-18a-3p MIMAT0021818
          Chicken gga-miR-18a-3p MIMAT0026499
          Chrysemys picta cpi-miR-18a-3p MIMAT0037639
          Ophiophagus hannah oha-miR-18a-3p MIMAT0036769
          View More Rows
          Python bivittatus pbv-miR-18a-3p MIMAT0038841

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-18a
          Stem-loop Accession # MI0000567
          Stem-loop Sequence
          UGCGUGCUUUUUGUUCUAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGACUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGCAUAAGAAGUUAUGUC
          Chromosome Location Chr. 14 - 115043851 - 115043946 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004626
          miRBase ID: mmu-miR-18a-3p
          Mature miRNA Sequence: ACUGCCCUAAGUGCUCCUUCUG
          Chromosome Location: Chr. 14 - 115043851 - 115043946 [+] on Build GRCm38

          Alligator mississippiensis

          Stem-loop ID ami-mir-18
          Stem-loop Accession # MI0029673
          Stem-loop Sequence
          GCUUUUUGUUCUAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGCAUAAGAAGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038050
          miRBase ID: ami-miR-18-3p
          Mature miRNA Sequence: ACUGCCCUAAGUGCUCCUUCUG
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-18a
          Stem-loop Accession # MI0018783
          Stem-loop Sequence
          AGUGACUGCUUUUUGUUCUAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGCAUAAGAAGUUGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021818
          miRBase ID: aca-miR-18a-3p
          Mature miRNA Sequence: ACUGCCCUAAGUGCUCCUUCUG
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-18a
          Stem-loop Accession # MI0001183
          Stem-loop Sequence
          GUGCUUUUUGUACUAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGCAUAAGAAGUUAUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026499
          miRBase ID: gga-miR-18a-3p
          Mature miRNA Sequence: ACUGCCCUAAGUGCUCCUUCUG
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-18a
          Stem-loop Accession # MI0029402
          Stem-loop Sequence
          UGCUUUUUGUUCUAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGCAUAAGAAGUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037639
          miRBase ID: cpi-miR-18a-3p
          Mature miRNA Sequence: ACUGCCCUAAGUGCUCCUUCUG
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-18a
          Stem-loop Accession # MI0031381
          Stem-loop Sequence
          UUUUUGUUCUAAGGUGCAUCUAGUGCAGAUAGUGAAAUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGCAUAAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036769
          miRBase ID: oha-miR-18a-3p
          Mature miRNA Sequence: ACUGCCCUAAGUGCUCCUUCUG
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-18a
          Stem-loop Accession # MI0030162
          Stem-loop Sequence
          AUAACUGCUUUUUGUUCUAAGGUGCAUCUAGUGCAGAUAGUGAAGUAGAUUAGCAUCUACUGCCCUAAGUGCUCCUUCUGGCAUAAGAAGUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038841
          miRBase ID: pbv-miR-18a-3p
          Mature miRNA Sequence: ACUGCCCUAAGUGCUCCUUCUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm04228426_pri
          Ambion® Pre-miR™ miRNA Precursor : PM12283
          TaqMan™ MicroRNA Assay : 002490
          mirVana® miRNA inhibitor : MH12283
          mirVana® miRNA mimic : MC12283
          TaqMan™ Advanced miRNA Assay : mmu481574_mir


          miRBase Alias:

          Mouse: mmu-miR-18a* (v17)
          Aeromonas caviae Ae398: aca-miR-18a* (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch