Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM12301
          Assay Name: hsa-miR-223-3p
          miRBase Accession Number: MI0000300
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 66018870 - 66018979 [+] on Build GRCh38
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCCA
          Species: Human, Mouse, Bovine, Callithrix jacchus, Chrysemys picta, Columba livia, Gadus morhua, Goat, Horse
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM12301
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000067, mir-223

          Mature miRNA Sequence:

          UGUCAGUUUGUCAAAUACCCCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-223-3p MIMAT0000280
          Mouse mmu-miR-223-3p MIMAT0000665
          Bovine bta-miR-223 MIMAT0009270
          Callithrix jacchus cja-miR-223 MIMAT0039506
          Chrysemys picta cpi-miR-223-3p MIMAT0037857
          Columba livia cli-miR-223-3p MIMAT0038610
          View More Rows
          Gadus morhua gmo-miR-223a-3p MIMAT0046041
          Goat chi-miR-223-3p MIMAT0036072
          Horse eca-miR-223 MIMAT0013205

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-223
          Stem-loop Accession # MI0000300
          Stem-loop Sequence
          CCUGGCCUCCUGCAGUGCCACGCUCCGUGUAUUUGACAAGCUGAGUUGGACACUCCAUGUGGUAGAGUGUCAGUUUGUCAAAUACCCCAAGUGCGGCACAUGCUUACCAG
          Chromosome Location Chr. X - 66018870 - 66018979 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000280
          miRBase ID: hsa-miR-223-3p
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCCA
          Chromosome Location: Chr. X - 66018870 - 66018979 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-223
          Stem-loop Accession # MI0000703
          Stem-loop Sequence
          UCUGGCCAUCUGCAGUGUCACGCUCCGUGUAUUUGACAAGCUGAGUUGGACACUCUGUGUGGUAGAGUGUCAGUUUGUCAAAUACCCCAAGUGUGGCUCAUGCCUAUCAG
          Chromosome Location Chr. X - 96242817 - 96242926 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000665
          miRBase ID: mmu-miR-223-3p
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCCA
          Chromosome Location: Chr. X - 96242817 - 96242926 [+] on Build GRCm38

          Bovine

          Stem-loop ID bta-mir-223
          Stem-loop Accession # MI0009782
          Stem-loop Sequence
          CCCAGCCUCCUGCAGUGCCAUGCUCCGUGUAUUUGACAAGCUGAGUUGGACACUCCAUGUAGUAGUGUCAGUUUGUCAAAUACCCCAAGUGUGGCAUAUGCCUAGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009270
          miRBase ID: bta-miR-223
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCCA
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-223
          Stem-loop Accession # MI0032032
          Stem-loop Sequence
          CCCUGGCCUGGCCUCCUGCAGUGCCACGCUCCGUGUAUUUGACAAGCUGAGUUAGACACUCCGUGUGGUAGAGUGUCAGUUUGUCAAAUACCCCAAGUGUGACACAUGCUUAGCAGCUCCAGACCAGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039506
          miRBase ID: cja-miR-223
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCCA
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-223
          Stem-loop Accession # MI0029549
          Stem-loop Sequence
          UGCAGUGCUGCGCUCCGUGUAUUUGACAAGCUGAGUUUGACACUCAAUGUGGCAGAGUGUCAGUUUGUCAAAUACCCCAAGUGAAGCAUUUGCUUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037857
          miRBase ID: cpi-miR-223-3p
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCCA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-223
          Stem-loop Accession # MI0030025
          Stem-loop Sequence
          AGCGCGGUGCCGCGCUGCGUGUAUUUGACAAGCUGAGUCCGACACUCAGUGCGAGAGUGUCAGUUUGUCAAAUACCCCAAGCGAGGCACUGCUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038610
          miRBase ID: cli-miR-223-3p
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCCA
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-223a
          Stem-loop Accession # MI0037988
          Stem-loop Sequence
          AGUGUAUUUGACAAGCUGAGCUGGACACUAAAAGACAGAGUGUCAGUUUGUCAAAUACCCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046041
          miRBase ID: gmo-miR-223a-3p
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCCA
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-223
          Stem-loop Accession # MI0030698
          Stem-loop Sequence
          CCAGCCUUCUGCAGUGCCAUGCUCUGUGUAUUUGACAAGCUGAGUUGGACACUCCAUGUAGUAGUGUCAGUUUGUCAAAUACCCCAAGUGCGGCAUAUGCCUAGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036072
          miRBase ID: chi-miR-223-3p
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCCA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-223
          Stem-loop Accession # MI0012953
          Stem-loop Sequence
          GCCCAGCCUCCUGCAGUGCCACGCUCCGUGUAUUUGACAAGCUGAGUUGGACACUCCAUGUGGUAGAGUGUCAGUUUGUCAAAUACCCCAAGUGCGGCACAUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013205
          miRBase ID: eca-miR-223
          Mature miRNA Sequence: UGUCAGUUUGUCAAAUACCCCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303017_pri
          TaqMan™ Pri-miRNA Assay : Mm03307156_pri
          Ambion® Pre-miR™ miRNA Precursor : PM12301
          TaqMan™ MicroRNA Assay : 002295
          mirVana® miRNA inhibitor : MH12301
          mirVana® miRNA mimic : MC12301
          TaqMan™ Advanced miRNA Assay : 477983_mir
          TaqMan™ Advanced miRNA Assay : mmu481007_mir


          miRBase Alias:

          Human: hsa-miR-223 (v17)
          Mouse: mmu-miR-223 (v17)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b6cf8ccb7-57dk8:80/100.66.72.150:80.
          git-commit: 76b2fce788d0185f5e22c122d85c407206860d42
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.0-2026-offline-prod