Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM10492
          Assay Name: hsa-miR-200b-3p
          miRBase Accession Number: MI0000342
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 1167104 - 1167198 [+] on Build GRCh38
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGA
          Species: Human, Mouse, Anolis carolinensis, Ciona savignyi, Cyprinus carpio, Fugu rubripes, Goat, Horse, Monodelphis domestica, Tetraodon nigroviridis, Tupaia chinensis, Zebrafish
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM10492
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000019, mir-8

          Mature miRNA Sequence:

          UAAUACUGCCUGGUAAUGAUGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-200b-3p MIMAT0000318
          Mouse mmu-miR-200b-3p MIMAT0000233
          Anolis carolinensis aca-miR-200b-3p MIMAT0021847
          Ciona savignyi csa-miR-200 MIMAT0006133
          Cyprinus carpio ccr-miR-200b MIMAT0026259
          Fugu rubripes fru-miR-200b MIMAT0002983
          View More Rows
          Goat chi-miR-200b MIMAT0036046
          Horse eca-miR-200b MIMAT0012910
          Monodelphis domestica mdo-miR-200b-3p MIMAT0004156
          Tetraodon nigroviridis tni-miR-200b MIMAT0002984
          Tupaia chinensis tch-miR-200b-3p MIMAT0036612
          Zebrafish dre-miR-200b-3p MIMAT0001862

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-200b
          Stem-loop Accession # MI0000342
          Stem-loop Sequence
          CCAGCUCGGGCAGCCGUGGCCAUCUUACUGGGCAGCAUUGGAUGGAGUCAGGUCUCUAAUACUGCCUGGUAAUGAUGACGGCGGAGCCCUGCACG
          Chromosome Location Chr. 1 - 1167104 - 1167198 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000318
          miRBase ID: hsa-miR-200b-3p
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGA
          Chromosome Location: Chr. 1 - 1167104 - 1167198 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-200b
          Stem-loop Accession # MI0000243
          Stem-loop Sequence
          GCCGUGGCCAUCUUACUGGGCAGCAUUGGAUAGUGUCUGAUCUCUAAUACUGCCUGGUAAUGAUGACGGC
          Chromosome Location Chr. 4 - 156055681 - 156055750 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000233
          miRBase ID: mmu-miR-200b-3p
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGA
          Chromosome Location: Chr. 4 - 156055681 - 156055750 [-] on Build GRCm38

          Anolis carolinensis

          Stem-loop ID aca-mir-200b
          Stem-loop Accession # MI0018803
          Stem-loop Sequence
          AAACCAUCUUGGGAGGUCAUUGCCAUCUUACUGGGCAGCAUUGGAUGUUUUCUGUCUUUCUAAUACUGCCUGGUAAUGAUGAUUGUGGCCUUCUCACGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021847
          miRBase ID: aca-miR-200b-3p
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGA
          Chromosome Location: NA

          Ciona savignyi

          Stem-loop ID csa-mir-200
          Stem-loop Accession # MI0007200
          Stem-loop Sequence
          GGUCACUGAAGCAUCAACCCGGUAGUACGAUAUGUGAGCUGUAUAAUACUGCCUGGUAAUGAUGAUUCAGUGGCGUCGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006133
          miRBase ID: csa-miR-200
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGA
          Chromosome Location: NA

          Cyprinus carpio

          Stem-loop ID ccr-mir-200b
          Stem-loop Accession # MI0023362
          Stem-loop Sequence
          UGGGUAGUCUUCUCCAUCUUACGAGGCAGCAUUGGAUUUCAUUAUUUUCUCUAAUACUGCCUGGUAAUGAUGAUGAUGGCUGUCCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026259
          miRBase ID: ccr-miR-200b
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGA
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-200b
          Stem-loop Accession # MI0003305
          Stem-loop Sequence
          GGUGAUUAUCUCCAUCUUACGAGGCAGCAUUGGAUAUCAUCACUUUCUCUAAUACUGCCUGGUAAUGAUGAUGAUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002983
          miRBase ID: fru-miR-200b
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGA
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-200b
          Stem-loop Accession # MI0030680
          Stem-loop Sequence
          GCCAUCGCCAUCUUACUGGGCAGCAUUGGAUGGUGUCUGGUCUCUAAUACUGCCUGGUAAUGAUGACGCUGGGGCCCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036046
          miRBase ID: chi-miR-200b
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-200b
          Stem-loop Accession # MI0012666
          Stem-loop Sequence
          GCCGUCGCCAUCUUACUGGGCAGCAUUGGAUGUGUCUGGUCUCUAAUACUGCCUGGUAAUGAUGACGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012910
          miRBase ID: eca-miR-200b
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGA
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-200b
          Stem-loop Accession # MI0005345
          Stem-loop Sequence
          CCAUCUUACUGGGCAGCAUUGGAUGGUGUCUGUGUUUCUAAUACUGCCUGGUAAUGAUGAUGAUGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004156
          miRBase ID: mdo-miR-200b-3p
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGA
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-200b
          Stem-loop Accession # MI0003306
          Stem-loop Sequence
          CCAUCUUACGAGGCAGCAUUGGAUAGCAUCACUUUUUCUAAUACUGCCUGGUAAUGAUGAUGAUCGUCGUCUGCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002984
          miRBase ID: tni-miR-200b
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGA
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-200b
          Stem-loop Accession # MI0031282
          Stem-loop Sequence
          CAUCUUACUGGGCAGCAUUGGAUGGUGUCUGGUCUCUAAUACUGCCUGGUAAUGAUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036612
          miRBase ID: tch-miR-200b-3p
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGA
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-200b
          Stem-loop Accession # MI0002038
          Stem-loop Sequence
          GGUAGUCGUCUCCAUCUUACGAGGCAGCAUUGGAUUUCAUUACUUUUUCUAAUACUGCCUGGUAAUGAUGAUGAUUGCUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001862
          miRBase ID: dre-miR-200b-3p
          Mature miRNA Sequence: UAAUACUGCCUGGUAAUGAUGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303027_pri
          TaqMan™ Pri-miRNA Assay : Mm03306509_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10492
          TaqMan™ MicroRNA Assay : 002251
          mirVana® miRNA inhibitor : MH10492
          mirVana® miRNA mimic : MC10492
          TaqMan™ Advanced miRNA Assay : 477963_mir
          TaqMan™ Advanced miRNA Assay : mmu482918_mir


          miRBase Alias:

          Human: hsa-miR-200b (v17)
          Mouse: mmu-miR-200b (v17)
          Aeromonas caviae Ae398: aca-miR-200b (v18)
          Danio rerio: dre-miR-200b (v20)
          Monodelphis domestica: mdo-miR-200b (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
          git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.49.0-Offline