Search
Search

| Assay Name: | hsa-miR-130a-3p |
|---|---|
| miRBase Accession Number: | MI0000448 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGUGCAAUGUUAAAAGGGCAU |
| Species: | Human, Mouse, Rat, Bovine, Chicken, Columba livia, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Dog, Guinea pig, Horse, Monodelphis domestica, Ophiophagus hannah, Ornithorhynchus anatinus, Otolemur garnettii, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Python bivittatus, Taeniopygia guttata, Tupaia chinensis, Xenopus tropicalis, Zebrafish |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000034, mir-130 |
Mature miRNA Sequence: | CAGUGCAAUGUUAAAAGGGCAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-130a-3p | MIMAT0000425 |
| Mouse | mmu-miR-130a-3p | MIMAT0000141 |
| Rat | rno-miR-130a-3p | MIMAT0000836 |
| Bovine | bta-miR-130a | MIMAT0009223 |
| Chicken | gga-miR-130c-3p | MIMAT0007734 |
| Columba livia | cli-miR-130a-3p | MIMAT0038506 |
| View More Rows | ||
| Cricetulus griseus | cgr-miR-130a-3p | MIMAT0023755 |
| Cyprinus carpio | ccr-miR-130a | MIMAT0026210 |
| Dasypus novemcinctus | dno-miR-130a-3p | MIMAT0047628 |
| Dog | cfa-miR-130a | MIMAT0006631 |
| Guinea pig | cpo-miR-130a-3p | MIMAT0046981 |
| Horse | eca-miR-130a | MIMAT0013047 |
| Monodelphis domestica | mdo-miR-130c-3p | MIMAT0028515 |
| Ophiophagus hannah | oha-miR-130b-3p | MIMAT0036690 |
| Ornithorhynchus anatinus | oan-miR-130b-3p | MIMAT0007159 |
| Otolemur garnettii | oga-miR-130a | MIMAT0049666 |
| Pan troglodytes | ptr-miR-130a | MIMAT0008025 |
| Pig | ssc-miR-130a | MIMAT0007758 |
| Pongo pygmaeus | ppy-miR-130a | MIMAT0015753 |
| Pteropus alecto | pal-miR-130-3p | MIMAT0039896 |
| Python bivittatus | pbv-miR-130d-3p | MIMAT0038932 |
| Taeniopygia guttata | tgu-miR-130c-3p | MIMAT0014581 |
| Tupaia chinensis | tch-miR-130a-3p | MIMAT0036577 |
| Xenopus tropicalis | xtr-miR-130a | MIMAT0003591 |
| Zebrafish | dre-miR-130a | MIMAT0001826 |