Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM10513
          Assay Name: hsa-miR-137-3p
          miRBase Accession Number: MI0000454
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 98046070 - 98046171 [-] on Build GRCh38
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Species: Human, Mouse, Rat, Anolis carolinensis, Bovine, Callithrix jacchus, Chrysemys picta, Columba livia, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Horse, Ictalurus punctatus, Oreochromis niloticus, Ornithorhynchus anatinus, Pan troglodytes, Pig, Pongo pygmaeus, Python bivittatus, Rhesus monkey, Taeniopygia guttata
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM10513
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000106, mir-137

          Mature miRNA Sequence:

          UUAUUGCUUAAGAAUACGCGUAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-137-3p MIMAT0000429
          Mouse mmu-miR-137-3p MIMAT0000149
          Rat rno-miR-137-3p MIMAT0000843
          Anolis carolinensis aca-miR-137a MIMAT0021754
          Bovine bta-miR-137 MIMAT0009231
          Callithrix jacchus cja-miR-137 MIMAT0039514
          View More Rows
          Chrysemys picta cpi-miR-137a-3p MIMAT0037758
          Columba livia cli-miR-137a-3p MIMAT0038519
          Cricetulus griseus cgr-miR-137-3p MIMAT0023763
          Cyprinus carpio ccr-miR-137 MIMAT0026217
          Dasypus novemcinctus dno-miR-137-3p MIMAT0047647
          Horse eca-miR-137 MIMAT0012953
          Ictalurus punctatus ipu-miR-137 MIMAT0029413
          Oreochromis niloticus oni-miR-137a MIMAT0042764
          Ornithorhynchus anatinus oan-miR-137a-3p MIMAT0006838
          Pan troglodytes ptr-miR-137 MIMAT0008033
          Pig ssc-miR-137 MIMAT0025362
          Pongo pygmaeus ppy-miR-137 MIMAT0015759
          Python bivittatus pbv-miR-137a-3p MIMAT0038946
          Rhesus monkey mml-miR-137-3p MIMAT0006193
          Taeniopygia guttata tgu-miR-137-3p MIMAT0014543

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-137
          Stem-loop Accession # MI0000454
          Stem-loop Sequence
          GGUCCUCUGACUCUCUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGAGUACCAGCGGCA
          Chromosome Location Chr. 1 - 98046070 - 98046171 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000429
          miRBase ID: hsa-miR-137-3p
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: Chr. 1 - 98046070 - 98046171 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-137
          Stem-loop Accession # MI0000163
          Stem-loop Sequence
          CUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGG
          Chromosome Location Chr. 3 - 118433857 - 118433929 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000149
          miRBase ID: mmu-miR-137-3p
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: Chr. 3 - 118433857 - 118433929 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-137
          Stem-loop Accession # MI0000910
          Stem-loop Sequence
          GGCCCUCUGACUCUCUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGAGUACCAGCGGCA
          Chromosome Location Chr. 2 - 221658106 - 221658207 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000843
          miRBase ID: rno-miR-137-3p
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: Chr. 2 - 221658106 - 221658207 [+] on Build Rnor_6.0

          Anolis carolinensis

          Stem-loop ID aca-mir-137a
          Stem-loop Accession # MI0018744
          Stem-loop Sequence
          AGCUUCCUGACUCUCUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGAGUACUCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021754
          miRBase ID: aca-miR-137a
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-137
          Stem-loop Accession # MI0009740
          Stem-loop Sequence
          GGUCCUCUGACUCUCUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGAGUACCAGCGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009231
          miRBase ID: bta-miR-137
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-137
          Stem-loop Accession # MI0032041
          Stem-loop Sequence
          ACUCUCUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039514
          miRBase ID: cja-miR-137
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-137a
          Stem-loop Accession # MI0029479
          Stem-loop Sequence
          ACUCUCUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGAGUACCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037758
          miRBase ID: cpi-miR-137a-3p
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-137a
          Stem-loop Accession # MI0029971
          Stem-loop Sequence
          CUGACUCUCUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGAGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038519
          miRBase ID: cli-miR-137a-3p
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-137
          Stem-loop Accession # MI0020399
          Stem-loop Sequence
          CUCUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023763
          miRBase ID: cgr-miR-137-3p
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: NA

          Cyprinus carpio

          Stem-loop ID ccr-mir-137
          Stem-loop Accession # MI0023323
          Stem-loop Sequence
          UAAAAGACUCUCUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGCUCUCGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGAGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026217
          miRBase ID: ccr-miR-137
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-137
          Stem-loop Accession # MI0038985
          Stem-loop Sequence
          ACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047647
          miRBase ID: dno-miR-137-3p
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-137
          Stem-loop Accession # MI0012705
          Stem-loop Sequence
          CUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012953
          miRBase ID: eca-miR-137
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-137
          Stem-loop Accession # MI0024500
          Stem-loop Sequence
          ACGGGUAUUCUUGGGUGGAUAAUACGGCUCUCGUUGUUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029413
          miRBase ID: ipu-miR-137
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-137a
          Stem-loop Accession # MI0034687
          Stem-loop Sequence
          ACGGGUAUUCUUGGGUGGAUAAUACAGAUCACGUUGUUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042764
          miRBase ID: oni-miR-137a
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-137a
          Stem-loop Accession # MI0006687
          Stem-loop Sequence
          GCUUCGUCUUUCUGACUCUCUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGAGUAUCUGCGGCAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006838
          miRBase ID: oan-miR-137a-3p
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-137
          Stem-loop Accession # MI0008538
          Stem-loop Sequence
          GGUCCUCUGACUCUCUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGAGUACCAGCGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008033
          miRBase ID: ptr-miR-137
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-137
          Stem-loop Accession # MI0022130
          Stem-loop Sequence
          ACUCUCUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025362
          miRBase ID: ssc-miR-137
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-137
          Stem-loop Accession # MI0014829
          Stem-loop Sequence
          GGUCCUCUGACUCUCUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGAGUACCAGCGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015759
          miRBase ID: ppy-miR-137
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-137a
          Stem-loop Accession # MI0030235
          Stem-loop Sequence
          AGCUUUCCGACUCUCUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGAGUACUCGCGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038946
          miRBase ID: pbv-miR-137a-3p
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-137
          Stem-loop Accession # MI0007627
          Stem-loop Sequence
          GGUCCUCUGACUCUCUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGAGUACCAGCGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006193
          miRBase ID: mml-miR-137-3p
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-137
          Stem-loop Accession # MI0013757
          Stem-loop Sequence
          GGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014543
          miRBase ID: tgu-miR-137-3p
          Mature miRNA Sequence: UUAUUGCUUAAGAAUACGCGUAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303136_pri
          TaqMan™ Pri-miRNA Assay : Rn03464604_pri
          TaqMan™ Pri-miRNA Assay : Mm03306305_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10513
          TaqMan™ MicroRNA Assay : 001129
          mirVana® miRNA inhibitor : MH10513
          mirVana® miRNA mimic : MC10513
          TaqMan™ Advanced miRNA Assay : rno480924_mir
          TaqMan™ Advanced miRNA Assay : mmu480924_mir
          TaqMan™ Advanced miRNA Assay : 477904_mir


          miRBase Alias:

          Human: hsa-miR-137 (v21)
          Mouse: mmu-miR-137 (v17)
          Rat: rno-miR-137 (v18)
          Macaca mulatta: mml-miR-137 (v19)
          Ochrobactrum anthropi: oan-miR-137a (v18)
          Trabulsiella guamensis ATCC 49490: tgu-miR-137 (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-8cpz7:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch