Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM11553
          Assay Name: hsa-miR-532-5p
          miRBase Accession Number: MI0003205
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 50003148 - 50003238 [+] on Build GRCh38
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACCGU
          Species: Human, Mouse, Bovine, Callithrix jacchus, Cricetulus griseus, Dog, Goat, Guinea pig, Horse, Microcebus murinus, Nomascus leucogenys, Papio hamadryas, Pig, Pongo pygmaeus, Rhesus monkey, Tupaia chinensis
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM11553
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000113, mir-188

          Mature miRNA Sequence:

          CAUGCCUUGAGUGUAGGACCGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-532-5p MIMAT0002888
          Mouse mmu-miR-532-5p MIMAT0002889
          Bovine bta-miR-532 MIMAT0003848
          Callithrix jacchus cja-miR-532 MIMAT0039399
          Cricetulus griseus cgr-miR-532-5p MIMAT0023981
          Dog cfa-miR-532 MIMAT0006758
          View More Rows
          Goat chi-miR-532-5p MIMAT0036270
          Guinea pig cpo-miR-532-5p MIMAT0047335
          Horse eca-miR-532-5p MIMAT0013235
          Microcebus murinus mmr-miR-532 MIMAT0049158
          Nomascus leucogenys nle-miR-532 MIMAT0049364
          Papio hamadryas pha-miR-532 MIMAT0049565
          Pig ssc-miR-532-5p MIMAT0013940
          Pongo pygmaeus ppy-miR-532-5p MIMAT0015987
          Rhesus monkey mml-miR-532-5p MIMAT0006403
          Tupaia chinensis tch-miR-532-5p MIMAT0036591

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-532
          Stem-loop Accession # MI0003205
          Stem-loop Sequence
          CGACUUGCUUUCUCUCCUCCAUGCCUUGAGUGUAGGACCGUUGGCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCAGAAGAGCGAGCCU
          Chromosome Location Chr. X - 50003148 - 50003238 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002888
          miRBase ID: hsa-miR-532-5p
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACCGU
          Chromosome Location: Chr. X - 50003148 - 50003238 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-532
          Stem-loop Accession # MI0003206
          Stem-loop Sequence
          CAGAUUUGCUUUUUCUCUUCCAUGCCUUGAGUGUAGGACCGUUGACAUCUUAAUUACCCUCCCACACCCAAGGCUUGCAGGAGAGCAAGCCUUCUC
          Chromosome Location Chr. X - 7248402 - 7248497 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0002889
          miRBase ID: mmu-miR-532-5p
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACCGU
          Chromosome Location: Chr. X - 7248402 - 7248497 [-] on Build GRCm38

          Bovine

          Stem-loop ID bta-mir-532
          Stem-loop Accession # MI0005061
          Stem-loop Sequence
          GACUUGCUUUCUCUCUUACAUGCCUUGAGUGUAGGACCGUUGGCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCAGGAGAGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003848
          miRBase ID: bta-miR-532
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACCGU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-532
          Stem-loop Accession # MI0031918
          Stem-loop Sequence
          GCUUUCUCUCCUCCAUGCCUUGAGUGUAGGACCGUUGGCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCAGAAGAGCGAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039399
          miRBase ID: cja-miR-532
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACCGU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-532
          Stem-loop Accession # MI0020539
          Stem-loop Sequence
          UUUCUCUUUCAUGCCUUGAGUGUAGGACCGUUAACAUCCUAAUUACCCUCCCACACCCAAGGUUUGCAGGAGAGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023981
          miRBase ID: cgr-miR-532-5p
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACCGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-532
          Stem-loop Accession # MI0008184
          Stem-loop Sequence
          CAUGCCUUGAGUGUAGGACCGUUGGUAUCUUAAUUACCCUCCCACACCCAAGGCUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006758
          miRBase ID: cfa-miR-532
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACCGU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-532
          Stem-loop Accession # MI0030818
          Stem-loop Sequence
          GACUUGCUUUCUCUCUUACAUGCCUUGAGUGUAGGACCGUUGGCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCAGGAGGGCCAGCCUUUUCCUCCCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036270
          miRBase ID: chi-miR-532-5p
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACCGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-532
          Stem-loop Accession # MI0038817
          Stem-loop Sequence
          CAUGCCUUGAGUGUAGGACCGUUGGCGUCUUAAUUACCCUCCCACACCCAAGGCUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047335
          miRBase ID: cpo-miR-532-5p
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACCGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-532
          Stem-loop Accession # MI0012979
          Stem-loop Sequence
          GAUUUGCUUUCUCUCUUACAUGCCUUGAGUGUAGGACCGUUGGCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCAGGAGGGCAAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013235
          miRBase ID: eca-miR-532-5p
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACCGU
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-532
          Stem-loop Accession # MI0039873
          Stem-loop Sequence
          CAUGCCUUGAGUGUAGGACCGUUGGCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049158
          miRBase ID: mmr-miR-532
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACCGU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-532
          Stem-loop Accession # MI0040101
          Stem-loop Sequence
          CAUGCCUUGAGUGUAGGACCGUUGGCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049364
          miRBase ID: nle-miR-532
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACCGU
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-532
          Stem-loop Accession # MI0040335
          Stem-loop Sequence
          CAUGCCUUGAGUGUAGGACCGUUGGCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049565
          miRBase ID: pha-miR-532
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACCGU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-532
          Stem-loop Accession # MI0013150
          Stem-loop Sequence
          UUGCUUUCUCUCUUACAUGCCUUGAGUGUAGGACCGUUGGCAUCUGAAUACCCUCCCACACCCAAGGCUUGCAGGAGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013940
          miRBase ID: ssc-miR-532-5p
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACCGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-532
          Stem-loop Accession # MI0015037
          Stem-loop Sequence
          CGACUUGCUUUCUCUCCUCCAUGCCUUGAGUGUAGGACCGUUGGCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCAGAAGAGCGAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015987
          miRBase ID: ppy-miR-532-5p
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACCGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-532
          Stem-loop Accession # MI0007814
          Stem-loop Sequence
          CGACUUGCUUUCUCUCCUCCAUGCCUUGAGUGUAGGACCGUUGGCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006403
          miRBase ID: mml-miR-532-5p
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACCGU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-532
          Stem-loop Accession # MI0031257
          Stem-loop Sequence
          CAUGCCUUGAGUGUAGGACCGUUGGCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036591
          miRBase ID: tch-miR-532-5p
          Mature miRNA Sequence: CAUGCCUUGAGUGUAGGACCGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03304216_pri
          TaqMan™ Pri-miRNA Assay : Mm03307612_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11553
          TaqMan™ MicroRNA Assay : 001518
          mirVana® miRNA inhibitor : MH11553
          mirVana® miRNA mimic : MC11553
          TaqMan™ Advanced miRNA Assay : 478151_mir


          miRBase Alias:

          Human: hsa-miR-532 (v9.2)
          Mouse: mmu-miR-532 (v9.2)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch