Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM12731
          Assay Name: hsa-miR-193b-5p
          miRBase Accession Number: MI0003137
          miRBase Version: v22.1
          Chromosome Location: Chr.16: 14303967 - 14304049 [+] on Build GRCh38
          Mature miRNA Sequence: CGGGGUUUUGAGGGCGAGAUGA
          Species: Human, Mouse, Cricetulus griseus, Dasypus novemcinctus, Dog, Goat, Gorilla gorilla, Guinea pig, Monodelphis domestica, Ornithorhynchus anatinus, Pteropus alecto, Rabbit, Rhesus monkey
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM12731
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000082, mir-193

          Mature miRNA Sequence:

          CGGGGUUUUGAGGGCGAGAUGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-193b-5p MIMAT0004767
          Mouse mmu-miR-193b-5p MIMAT0017271
          Cricetulus griseus cgr-miR-193b-5p MIMAT0023825
          Dasypus novemcinctus dno-miR-193b-5p MIMAT0047723
          Dog cfa-miR-193b MIMAT0006699
          Goat chi-miR-193b-5p MIMAT0036027
          View More Rows
          Gorilla gorilla ggo-miR-193b MIMAT0024138
          Guinea pig cpo-miR-193b-5p MIMAT0047072
          Monodelphis domestica mdo-miR-193b-5p MIMAT0028652
          Ornithorhynchus anatinus oan-miR-193-5p MIMAT0006820
          Pteropus alecto pal-miR-193b-5p MIMAT0039967
          Rabbit ocu-miR-193b-5p MIMAT0048305
          Rhesus monkey mml-miR-193b-5p MIMAT0026836

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-193b
          Stem-loop Accession # MI0003137
          Stem-loop Sequence
          GUGGUCUCAGAAUCGGGGUUUUGAGGGCGAGAUGAGUUUAUGUUUUAUCCAACUGGCCCUCAAAGUCCCGCUUUUGGGGUCAU
          Chromosome Location Chr. 16 - 14303967 - 14304049 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004767
          miRBase ID: hsa-miR-193b-5p
          Mature miRNA Sequence: CGGGGUUUUGAGGGCGAGAUGA
          Chromosome Location: Chr. 16 - 14303967 - 14304049 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-193b
          Stem-loop Accession # MI0005484
          Stem-loop Sequence
          GGCCCAGAAUCGGGGUUUUGAGGGCGAGAUGAGUUUGUGUUUUAUCCAACUGGCCCACAAAGUCCCGCUUUUGGGGUCA
          Chromosome Location Chr. 16 - 13449523 - 13449601 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017271
          miRBase ID: mmu-miR-193b-5p
          Mature miRNA Sequence: CGGGGUUUUGAGGGCGAGAUGA
          Chromosome Location: Chr. 16 - 13449523 - 13449601 [+] on Build GRCm38

          Cricetulus griseus

          Stem-loop ID cgr-mir-193b
          Stem-loop Accession # MI0020439
          Stem-loop Sequence
          GCCCAGAAUCGGGGUUUUGAGGGCGAGAUGAGUUUGUGUUUUAUCCAACUGGCCCACAAAGUCCCGCUUUUGGGGUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023825
          miRBase ID: cgr-miR-193b-5p
          Mature miRNA Sequence: CGGGGUUUUGAGGGCGAGAUGA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-193b
          Stem-loop Accession # MI0039025
          Stem-loop Sequence
          CGGGGUUUUGAGGGCGAGAUGAGUUUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047723
          miRBase ID: dno-miR-193b-5p
          Mature miRNA Sequence: CGGGGUUUUGAGGGCGAGAUGA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-193b
          Stem-loop Accession # MI0008113
          Stem-loop Sequence
          CGGGGUUUUGAGGGCGAGAUGAGUUUAUGUUUGAUCCAACUGGCCCACAAAGUCCCGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006699
          miRBase ID: cfa-miR-193b
          Mature miRNA Sequence: CGGGGUUUUGAGGGCGAGAUGA
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-193b
          Stem-loop Accession # MI0030667
          Stem-loop Sequence
          GGAGGCUGUGGUCCCAGAAUCGGGGUUUUGAGGGCGAGAUGAGUUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCUUUUGGGGUCAUUCUAGACGGCGAGGGAUUCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036027
          miRBase ID: chi-miR-193b-5p
          Mature miRNA Sequence: CGGGGUUUUGAGGGCGAGAUGA
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-193b
          Stem-loop Accession # MI0020685
          Stem-loop Sequence
          GGAGGUUGUGGUCUCAGAAUCGGGGUUUUGAGGGCGAGAUGAGUUUAUGUUUUAUCCAACUGGCCCUCAAAGUCCCGCUUUUGGGGUCAUUCUGGACAGCGAGGGACUCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024138
          miRBase ID: ggo-miR-193b
          Mature miRNA Sequence: CGGGGUUUUGAGGGCGAGAUGA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-193b
          Stem-loop Accession # MI0038682
          Stem-loop Sequence
          CGGGGUUUUGAGGGCGAGAUGAGUUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047072
          miRBase ID: cpo-miR-193b-5p
          Mature miRNA Sequence: CGGGGUUUUGAGGGCGAGAUGA
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-193b
          Stem-loop Accession # MI0023850
          Stem-loop Sequence
          CGGGGUUUUGAGGGCGAGAUGAGUUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0028652
          miRBase ID: mdo-miR-193b-5p
          Mature miRNA Sequence: CGGGGUUUUGAGGGCGAGAUGA
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-193
          Stem-loop Accession # MI0006676
          Stem-loop Sequence
          GGUUGUGAUUCCAGAGUCGGGGUUUUGAGGGCGAGAUGAGUUUGUGUUUUAUCCAACUGGCCCACAAAGUCCCGCUUUUGGUGGUCACUUUGAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006820
          miRBase ID: oan-miR-193-5p
          Mature miRNA Sequence: CGGGGUUUUGAGGGCGAGAUGA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-193b
          Stem-loop Accession # MI0032425
          Stem-loop Sequence
          CGGGGUUUUGAGGGCGAGAUGAGUUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039967
          miRBase ID: pal-miR-193b-5p
          Mature miRNA Sequence: CGGGGUUUUGAGGGCGAGAUGA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-193b
          Stem-loop Accession # MI0039331
          Stem-loop Sequence
          CGGGGUUUUGAGGGCGAGAUGAGUUUAUGUUUUAUCCAACUGGCCCACAAAGUCCCGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048305
          miRBase ID: ocu-miR-193b-5p
          Mature miRNA Sequence: CGGGGUUUUGAGGGCGAGAUGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-193b
          Stem-loop Accession # MI0007656
          Stem-loop Sequence
          GUGGUCUCAGAAUCGGGGUUUUGAGGGCGAGAUGAGUUUAUGUUUUAUCCAACUGGCCCUCAAAGUCCCGCUUUUGGGGUCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026836
          miRBase ID: mml-miR-193b-5p
          Mature miRNA Sequence: CGGGGUUUUGAGGGCGAGAUGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303897_pri
          TaqMan™ Pri-miRNA Assay : Mm03308440_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12731
          TaqMan™ MicroRNA Assay : 002366
          mirVana® miRNA inhibitor : MH12731
          mirVana® miRNA mimic : MC12731
          TaqMan™ Advanced miRNA Assay : mmu481503_mir
          TaqMan™ Advanced miRNA Assay : 478742_mir


          miRBase Alias:

          Human: hsa-miR-193b* (v17)
          Mouse: mmu-miR-193b* (v17)
          Ochrobactrum anthropi: oan-miR-193* (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch