Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM12824
          Assay Name: hsa-miR-532-3p
          miRBase Accession Number: MI0003205
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 50003148 - 50003238 [+] on Build GRCh38
          Mature miRNA Sequence: CCUCCCACACCCAAGGCUUGCA
          Species: Human, Mouse, Rat, Dasypus novemcinctus, Guinea pig, Horse, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM12824
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000113, mir-188

          Mature miRNA Sequence:

          CCUCCCACACCCAAGGCUUGCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-532-3p MIMAT0004780
          Mouse mmu-miR-532-3p MIMAT0004781
          Rat rno-miR-532-3p MIMAT0005323
          Dasypus novemcinctus dno-miR-532-3p MIMAT0047964
          Guinea pig cpo-miR-532-3p MIMAT0047336
          Horse eca-miR-532-3p MIMAT0013236
          View More Rows
          Pan troglodytes ptr-miR-532 MIMAT0008213
          Pig ssc-miR-532-3p MIMAT0013941
          Pongo pygmaeus ppy-miR-532-3p MIMAT0015988
          Pteropus alecto pal-miR-532-3p MIMAT0039902
          Rabbit ocu-miR-532-3p MIMAT0048520
          Rhesus monkey mml-miR-532-3p MIMAT0006404

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-532
          Stem-loop Accession # MI0003205
          Stem-loop Sequence
          CGACUUGCUUUCUCUCCUCCAUGCCUUGAGUGUAGGACCGUUGGCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCAGAAGAGCGAGCCU
          Chromosome Location Chr. X - 50003148 - 50003238 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004780
          miRBase ID: hsa-miR-532-3p
          Mature miRNA Sequence: CCUCCCACACCCAAGGCUUGCA
          Chromosome Location: Chr. X - 50003148 - 50003238 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-532
          Stem-loop Accession # MI0003206
          Stem-loop Sequence
          CAGAUUUGCUUUUUCUCUUCCAUGCCUUGAGUGUAGGACCGUUGACAUCUUAAUUACCCUCCCACACCCAAGGCUUGCAGGAGAGCAAGCCUUCUC
          Chromosome Location Chr. X - 7248402 - 7248497 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004781
          miRBase ID: mmu-miR-532-3p
          Mature miRNA Sequence: CCUCCCACACCCAAGGCUUGCA
          Chromosome Location: Chr. X - 7248402 - 7248497 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-532
          Stem-loop Accession # MI0006154
          Stem-loop Sequence
          UUUUCUCUUCCAUGCCUUGAGUGUAGGACUGUUGACAUCUUAAUUACCCUCCCACACCCAAGGCUUGCAGGAGAGCAAG
          Chromosome Location Chr. X - 16109870 - 16109948 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0005323
          miRBase ID: rno-miR-532-3p
          Mature miRNA Sequence: CCUCCCACACCCAAGGCUUGCA
          Chromosome Location: Chr. X - 16109870 - 16109948 [+] on Build Rnor_6.0

          Dasypus novemcinctus

          Stem-loop ID dno-mir-532
          Stem-loop Accession # MI0039148
          Stem-loop Sequence
          CAUGCCUUGAGUGUAGGACUGUGGGCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047964
          miRBase ID: dno-miR-532-3p
          Mature miRNA Sequence: CCUCCCACACCCAAGGCUUGCA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-532
          Stem-loop Accession # MI0038817
          Stem-loop Sequence
          CAUGCCUUGAGUGUAGGACCGUUGGCGUCUUAAUUACCCUCCCACACCCAAGGCUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047336
          miRBase ID: cpo-miR-532-3p
          Mature miRNA Sequence: CCUCCCACACCCAAGGCUUGCA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-532
          Stem-loop Accession # MI0012979
          Stem-loop Sequence
          GAUUUGCUUUCUCUCUUACAUGCCUUGAGUGUAGGACCGUUGGCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCAGGAGGGCAAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013236
          miRBase ID: eca-miR-532-3p
          Mature miRNA Sequence: CCUCCCACACCCAAGGCUUGCA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-532
          Stem-loop Accession # MI0008741
          Stem-loop Sequence
          GACUUGCUUUCUCUCCUCCAUGCCUUGAGUGUAGGACCGUUGGCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCAGAAGAGCGAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008213
          miRBase ID: ptr-miR-532
          Mature miRNA Sequence: CCUCCCACACCCAAGGCUUGCA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-532
          Stem-loop Accession # MI0013150
          Stem-loop Sequence
          UUGCUUUCUCUCUUACAUGCCUUGAGUGUAGGACCGUUGGCAUCUGAAUACCCUCCCACACCCAAGGCUUGCAGGAGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013941
          miRBase ID: ssc-miR-532-3p
          Mature miRNA Sequence: CCUCCCACACCCAAGGCUUGCA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-532
          Stem-loop Accession # MI0015037
          Stem-loop Sequence
          CGACUUGCUUUCUCUCCUCCAUGCCUUGAGUGUAGGACCGUUGGCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCAGAAGAGCGAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015988
          miRBase ID: ppy-miR-532-3p
          Mature miRNA Sequence: CCUCCCACACCCAAGGCUUGCA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-532
          Stem-loop Accession # MI0032391
          Stem-loop Sequence
          CAUGCCUUGAGUGUAGGACUGUUGCCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039902
          miRBase ID: pal-miR-532-3p
          Mature miRNA Sequence: CCUCCCACACCCAAGGCUUGCA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-532
          Stem-loop Accession # MI0039442
          Stem-loop Sequence
          CAUGCCUUGAGUGUAGGACUGUUGGAAUCUUAAUUACCCUCCCACACCCAAGGCUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048520
          miRBase ID: ocu-miR-532-3p
          Mature miRNA Sequence: CCUCCCACACCCAAGGCUUGCA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-532
          Stem-loop Accession # MI0007814
          Stem-loop Sequence
          CGACUUGCUUUCUCUCCUCCAUGCCUUGAGUGUAGGACCGUUGGCAUCUUAAUUACCCUCCCACACCCAAGGCUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006404
          miRBase ID: mml-miR-532-3p
          Mature miRNA Sequence: CCUCCCACACCCAAGGCUUGCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03304216_pri
          TaqMan™ Pri-miRNA Assay : Rn03465601_pri
          TaqMan™ Pri-miRNA Assay : Mm03307612_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12824
          TaqMan™ MicroRNA Assay : 002355
          mirVana® miRNA inhibitor : MH12824
          mirVana® miRNA mimic : MC12824
          TaqMan™ Advanced miRNA Assay : rno478336_mir
          TaqMan™ Advanced miRNA Assay : 478336_mir

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch