Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM15183
          Assay Name: hsa-miR-103a-2-5p
          miRBase Accession Number: MI0000108
          miRBase Version: v22.1
          Chromosome Location: Chr.20: 3917494 - 3917571 [+] on Build GRCh38
          Mature miRNA Sequence: AGCUUCUUUACAGUGCUGCCUUG
          Species: Human, Mouse, Alligator mississippiensis, Chicken, Columba livia, Guinea pig, Monodelphis domestica, Ophiophagus hannah, Ornithorhynchus anatinus, Rabbit
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM15183
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000024, mir-103

          Mature miRNA Sequence:

          AGCUUCUUUACAGUGCUGCCUUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-103a-2-5p MIMAT0009196
          Mouse mmu-miR-103-2-5p MIMAT0017025
          Alligator mississippiensis ami-miR-107-5p MIMAT0038121
          Chicken gga-miR-103-2-5p MIMAT0026519
          Columba livia cli-miR-103-5p MIMAT0038486
          Guinea pig cpo-miR-103-2-5p MIMAT0046952
          View More Rows
          Monodelphis domestica mdo-miR-103-2-5p MIMAT0031097
          Ophiophagus hannah oha-miR-103b-5p MIMAT0036658
          Ornithorhynchus anatinus oan-miR-103-2-5p MIMAT0006941
          Rabbit ocu-miR-103a-2-5p MIMAT0048191

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-103a-2
          Stem-loop Accession # MI0000108
          Stem-loop Sequence
          UUGUGCUUUCAGCUUCUUUACAGUGCUGCCUUGUAGCAUUCAGGUCAAGCAGCAUUGUACAGGGCUAUGAAAGAACCA
          Chromosome Location Chr. 20 - 3917494 - 3917571 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0009196
          miRBase ID: hsa-miR-103a-2-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGCUGCCUUG
          Chromosome Location: Chr. 20 - 3917494 - 3917571 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-103-2
          Stem-loop Accession # MI0000588
          Stem-loop Sequence
          GUCUUCGUGCUUUCAGCUUCUUUACAGUGCUGCCUUGUAGCAUUCAGGUCAAGCAGCAUUGUACAGGGCUAUGAAAGAACCAAGAA
          Chromosome Location Chr. 2 - 131288052 - 131288137 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017025
          miRBase ID: mmu-miR-103-2-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGCUGCCUUG
          Chromosome Location: Chr. 2 - 131288052 - 131288137 [+] on Build GRCm38

          Alligator mississippiensis

          Stem-loop ID ami-mir-107
          Stem-loop Accession # MI0029715
          Stem-loop Sequence
          GUAUUCUCUUUGCUUUCAGCUUCUUUACAGUGCUGCCUUGUUGCAUUUAUGUCAAGCAGCAUUGUACAGGGCUAUCAAAGCGGAGAGAGCUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038121
          miRBase ID: ami-miR-107-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGCUGCCUUG
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-103-2
          Stem-loop Accession # MI0001213
          Stem-loop Sequence
          UCUCUGUGCUUUCAGCUUCUUUACAGUGCUGCCUUGUUGCGUUCAUGUCAAGCAGCAUUGUACAGGGCUAUGAAAGAACAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026519
          miRBase ID: gga-miR-103-2-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGCUGCCUUG
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-103-2
          Stem-loop Accession # MI0029950
          Stem-loop Sequence
          UUAUCUUUGUGCUUUCAGCUUCUUUACAGUGCUGCCUUGUUGCGUUCAUGUCAAGCAGCAUUGUACAGGGCUAUGAAAGAGCAGAGACGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038486
          miRBase ID: cli-miR-103-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGCUGCCUUG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-103-2
          Stem-loop Accession # MI0038616
          Stem-loop Sequence
          AGCUUCUUUACAGUGCUGCCUUGUAGCAUUCAGGUCAAGCAGCAUUGUACAGGGCUAUGAAAGAACCAAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046952
          miRBase ID: cpo-miR-103-2-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGCUGCCUUG
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-103-2
          Stem-loop Accession # MI0005285
          Stem-loop Sequence
          CUUGGUGCUUUCAGCUUCUUUACAGUGCUGCCUUGUUGCAUUGAUGUCAAGCAGCAUUGUACAGGGCUAUGAAAGAACCAAGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031097
          miRBase ID: mdo-miR-103-2-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGCUGCCUUG
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-103b
          Stem-loop Accession # MI0031318
          Stem-loop Sequence
          CUGGGUGCUUCCAGCUUCUUUACAGUGCUGCCUUGUUGCAACCAUGUCAAGCAGCAUUGUACAGGGCUAUGAAAGACCCCAAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036658
          miRBase ID: oha-miR-103b-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGCUGCCUUG
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-103-2
          Stem-loop Accession # MI0006748
          Stem-loop Sequence
          GAGCUGCGCCGUCCUUGGUGCUUUCAGCUUCUUUACAGUGCUGCCUUGUUGCAUUCAUGUCAAGCAGCAUUGUACAGGGCUAUGAAAGAACCAAGGUUUUUGCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006941
          miRBase ID: oan-miR-103-2-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGCUGCCUUG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-103a-2
          Stem-loop Accession # MI0039269
          Stem-loop Sequence
          AGCUUCUUUACAGUGCUGCCUUGUAGCAUUCAGGUCAAGCAGCAUUGUACAGGGCUAUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048191
          miRBase ID: ocu-miR-103a-2-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGCUGCCUUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302758_pri
          TaqMan™ Pri-miRNA Assay : Mm03306917_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM15183
          TaqMan™ MicroRNA Assay : 121218_mat
          mirVana® miRNA inhibitor : MH15183
          mirVana® miRNA mimic : MC15183
          TaqMan™ Advanced miRNA Assay : 477864_mir


          miRBase Alias:

          Human: hsa-miR-103a-2* (v17)
          Human: hsa-miR-103-2* (v16)
          Mouse: mmu-miR-103-2* (v17)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch