Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
KCNE1
KCNE1B
Entrez Gene ID
3753
102723475
Gene Name:
potassium voltage-gated channel subfamily E regulatory subunit 1
potassium voltage-gated channel subfamily E regulatory subunit 1B
Gene Aliases
ISK, JLNS, JLNS2, LQT2/5, LQT5, MinK
-
Gene Chromosome Location
Chr.21: on Build GRCh38
Chr.21: on Build GRCh38
UniGene
Hs.121495
Species:
Human
Primer Chromosome Location
Chr.21: 7825266-7825444 on Build GRCh38
View in UCSC browser
Amplicon Length
179
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00476114_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
KCNE1
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_000219.5
NP_000210.2
-
-
-
NM_001127668.3
NP_001121140.1
-
-
-
NM_001127669.3
NP_001121141.1
-
-
-
NM_001127670.3
NP_001121142.1
-
-
-
NM_001270402.2
NP_001257331.1
-
-
-
NM_001270403.2
NP_001257332.1
-
-
-
NM_001270404.2
NP_001257333.1
-
-
-
NM_001270405.2
NP_001257334.1
-
-
-
XM_017028342.1
XP_016883831.1
-
-
-
GenBank
AK304336.1
-
-
-
AK311384.1
-
-
-
AK313543.1
-
-
-
AY789479.1
AAX11417.1
-
-
-
AY789480.1
AAX11418.1
-
-
-
BC036452.1
AAH36452.1
-
-
-
BC046224.1
AAH46224.1
-
-
-
DA418828.1
-
-
-
DA806205.1
-
-
-
DB089177.1
-
-
-
DB214219.1
-
-
-
DB223354.1
-
-
-
DB230009.1
-
-
-
DB233067.1
-
-
-
ENST00000399284.0
-
-
-
EU008568.1
-
-
-
EU008569.1
-
-
-
EU008570.1
-
-
-
Phenotype
MIM: 176261
KCNE1B
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
XM_006723946.2
XP_006724009.1
-
-
-
XM_006723949.2
XP_006724012.1
-
-
-
Phenotype
Literature Links:
KCNE1
PubMed Links
SNP ID
rs8133457
rs968435423
rs8133592
rs1020106766
rs910864554
rs978556582
rs988568695
rs868494281
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.21: 34455347-34455347 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.21: 34455351-34455351 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.21: 34455366-34455366 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.21: 34455334-34455334 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.21: 34455399-34455399 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.21: 34455367-34455367 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.21: 34455315-34455315 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Submission count
NA
Allele Nomenclature
Chromosome Location
Chr.21: 34455364-34455364 on Build GRCh38
Allele Frequency
1000Genome
Applied Biosystems
®
HapMap
Global - Not Available
Caucasian - Not Available
CEPH (CEU) - Not Available
EAS - Not Available
African American - Not Available
YRI (Yoruba) - Not Available
SAS - Not Available
Chinese - Not Available
CHB (Han Chinese) - Not Available
AFR - Not Available
Japanese - Not Available
JPT (Japanese) - Not Available
EUR - Not Available
AMR - Not Available
Back To Top
More Information
Set Membership:
Less than 200 bp
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
protein N-linked glycosylation
protein O-linked glycosylation
sensory perception of sound
regulation of ventricular cardiac muscle cell membrane repolarization
cardiac conduction
cellular response to cAMP
potassium ion export
potassium ion transmembrane transport
cardiac muscle cell action potential involved in contraction
ventricular cardiac muscle cell action potential
membrane repolarization
membrane repolarization during action potential
membrane repolarization during cardiac muscle cell action potential
regulation of heart rate by cardiac conduction
negative regulation of protein targeting to membrane
membrane repolarization during ventricular cardiac muscle cell action potential
regulation of potassium ion transmembrane transport
positive regulation of potassium ion transmembrane transport
regulation of delayed rectifier potassium channel activity
negative regulation of delayed rectifier potassium channel activity
voltage-gated potassium channel activity
delayed rectifier potassium channel activity
protein binding
potassium channel regulator activity
telethonin binding
voltage-gated potassium channel activity involved in cardiac muscle cell action potential repolarization
voltage-gated potassium channel activity involved in ventricular cardiac muscle cell action potential repolarization
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0