Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTNNB1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001098209.1 | NP_001091679.1 | - | - | - |
NM_001098210.1 | NP_001091680.1 | - | - | - | |
NM_001904.3 | NP_001895.1 | 1642257 | c.884C>A | p.A295D | |
5727 | c.860A>G | p.N287S | |||
1044594 | c.830G>A | p.G277D | |||
1044593 | c.777C>A | p.L259L | |||
188059 | c.770C>T | p.T257I | |||
1423045 | c.875A>C | p.K292T | |||
4441756 | c.848T>C | p.V283A | |||
4675396 | c.785delT | p.L263fs*13 | |||
1633163 | c.753G>T | p.V251V | |||
1423041 | c.767T>C | p.I256T | |||
6963932 | c.807_819>A | p.K270_V273delKMAV | |||
6949823 | c.803G>C | p.G268A | |||
5352212 | c.801A>T | p.E267D | |||
6345843 | c.821G>T | p.R274L | |||
4117547 | c.852C>T | p.A284A | |||
6854098 | c.821G>A | p.R274H | |||
3593970 | c.874A>G | p.K292E | |||
5608172 | c.737C>T | p.S246L | |||
2987955 | c.825A>T | p.L275F | |||
1186242 | c.883G>A | p.A295T | |||
7412938 | c.827C>G | p.A276G | |||
3696077 | c.768T>G | p.I256M | |||
4386454 | c.896A>T | p.D299V | |||
XM_005264886.2 | XP_005264943.1 | - | - | - | |
XM_006712983.1 | XP_006713046.1 | - | - | - | |
XM_006712984.1 | XP_006713047.1 | - | - | - | |
XM_006712985.1 | XP_006713048.1 | - | - | - | |
XM_017005738.1 | XP_016861227.1 | - | - | - | |
GenBank | AB062292.1 | BAB93475.1 | - | - | - |
AB451264.1 | - | - | - | ||
AB451392.1 | - | - | - | ||
AK289932.1 | - | - | - | ||
AK294778.1 | - | - | - | ||
AK296790.1 | - | - | - | ||
AK299954.1 | - | - | - | ||
AK314776.1 | - | - | - | ||
BC058926.1 | AAH58926.1 | - | - | - | |
ENST00000349496.0 | - | - | - | ||
X87838.1 | CAA61107.1 | - | - | - | |
Z19054.1 | CAA79497.1 | - | - | - |
Literature Links: |
CTNNB1 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |