Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FGFR1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001174063.1 | NP_001167534.1 | - | - | - |
NM_001174064.1 | NP_001167535.1 | 6918212 | c.272A>G | p.Y91C | |
3699075 | c.314A>G | p.Y105C | |||
1236024 | c.296C>T | p.S99L | |||
6953167 | c.287G>A | p.S96N | |||
6925252 | c.314A>T | p.Y105F | |||
6853425 | c.322G>A | p.V108I | |||
6925707 | c.280G>A | p.V94I | |||
3834686 | c.286A>C | p.S96R | |||
NM_001174065.1 | NP_001167536.1 | - | - | - | |
NM_001174066.1 | NP_001167537.1 | - | - | - | |
NM_001174067.1 | NP_001167538.1 | - | - | - | |
NM_015850.3 | NP_056934.2 | - | - | - | |
NM_023105.2 | NP_075593.1 | - | - | - | |
NM_023106.2 | NP_075594.1 | - | - | - | |
NM_023110.2 | NP_075598.2 | 1236023 | c.320C>T | p.S107L | |
3699074 | c.338A>G | p.Y113C | |||
6953166 | c.311G>A | p.S104N | |||
6918211 | c.296A>G | p.Y99C | |||
3699076 | c.338A>G | p.Y113C | |||
3834685 | c.310A>C | p.S104R | |||
6918213 | c.296A>G | p.Y99C | |||
6853424 | c.346G>A | p.V116I | |||
6925706 | c.304G>A | p.V102I | |||
6925251 | c.338A>T | p.Y113F | |||
6953168 | c.311G>A | p.S104N | |||
3834687 | c.310A>C | p.S104R | |||
6853426 | c.346G>A | p.V116I | |||
6925253 | c.338A>T | p.Y113F | |||
6925708 | c.304G>A | p.V102I | |||
5075011 | c.303C>T | p.C101C | |||
XM_006716303.2 | XP_006716366.1 | - | - | - | |
XM_006716304.1 | XP_006716367.1 | - | - | - | |
XM_006716306.2 | XP_006716369.1 | - | - | - | |
XM_006716307.1 | XP_006716370.1 | - | - | - | |
XM_006716309.3 | XP_006716372.1 | - | - | - | |
XM_006716310.2 | XP_006716373.1 | - | - | - | |
XM_006716311.1 | XP_006716374.1 | - | - | - | |
XM_006716312.1 | XP_006716375.1 | - | - | - | |
XM_006716313.2 | XP_006716376.1 | - | - | - | |
XM_006716314.1 | XP_006716377.1 | - | - | - | |
XM_011544443.2 | XP_011542745.1 | - | - | - | |
XM_011544444.1 | XP_011542746.1 | - | - | - | |
XM_011544445.2 | XP_011542747.1 | - | - | - | |
XM_011544446.2 | XP_011542748.1 | - | - | - | |
XM_011544447.2 | XP_011542749.1 | - | - | - | |
XM_011544448.1 | XP_011542750.1 | - | - | - | |
XM_011544449.1 | XP_011542751.1 | - | - | - | |
XM_011544450.2 | XP_011542752.1 | - | - | - | |
XM_011544451.1 | XP_011542753.1 | - | - | - | |
XM_011544452.2 | XP_011542754.1 | - | - | - | |
XM_017013219.1 | XP_016868708.1 | - | - | - | |
XM_017013220.1 | XP_016868709.1 | - | - | - | |
XM_017013221.1 | XP_016868710.1 | - | - | - | |
XM_017013222.1 | XP_016868711.1 | - | - | - | |
XM_017013223.1 | XP_016868712.1 | - | - | - | |
XM_017013224.1 | XP_016868713.1 | - | - | - | |
XM_017013225.1 | XP_016868714.1 | - | - | - | |
XM_017013226.1 | XP_016868715.1 | - | - | - | |
XM_017013227.1 | XP_016868716.1 | - | - | - | |
XM_017013228.1 | XP_016868717.1 | - | - | - | |
XM_017013230.1 | XP_016868719.1 | - | - | - | |
XM_017013231.1 | XP_016868720.1 | - | - | - | |
GenBank | AB208919.1 | BAD92156.1 | - | - | - |
AK222718.1 | BAD96438.1 | - | - | - | |
AK223126.1 | BAD96846.1 | - | - | - | |
AK291754.1 | - | - | - | ||
AK292470.1 | - | - | - | ||
AK309947.1 | - | - | - | ||
BC015035.1 | AAH15035.1 | - | - | - | |
BC018128.1 | AAH18128.1 | - | - | - | |
BC091494.1 | AAH91494.1 | - | - | - | |
BX649167.1 | - | - | - | ||
ENST00000447712.0 | - | - | - | ||
EU826596.1 | - | - | - | ||
EU826597.1 | - | - | - | ||
FJ809916.1 | - | - | - | ||
FJ809917.1 | - | - | - | ||
KU178066.1 | - | - | - | ||
KU178067.1 | - | - | - | ||
KU178068.1 | - | - | - | ||
KU178069.1 | - | - | - | ||
KU178070.1 | - | - | - | ||
M34185.1 | AAA35836.1 | - | - | - | |
M34186.1 | AAA35837.1 | - | - | - | |
M34187.1 | AAA35838.1 | - | - | - | |
M34188.1 | AAA35839.1 | - | - | - | |
M34641.1 | AAA35835.1 | - | - | - | |
M37722.1 | AAA75007.1 | - | - | - | |
M60485.1 | AAA35840.1 | - | - | - | |
M63887.1 | AAA35958.1 | - | - | - | |
M63888.1 | AAA35959.1 | - | - | - | |
M63889.1 | AAA35960.1 | - | - | - | |
S76658.1 | AAD14230.1 | - | - | - | |
X51803.1 | CAA36101.1 | - | - | - | |
X52833.1 | CAA37015.1 | - | - | - | |
X57118.1 | CAA40400.1 | - | - | - | |
X57119.1 | CAA40401.1 | - | - | - | |
X57120.1 | CAA40402.1 | - | - | - | |
X57121.1 | CAA40403.1 | - | - | - | |
X57122.1 | CAA40404.1 | - | - | - | |
X66945.1 | CAA47375.1 | - | - | - |
Literature Links: |
FGFR1 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |