Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ERG | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001136154.1 | NP_001129626.1 | - | - | - |
NM_001136155.1 | NP_001129627.1 | - | - | - | |
NM_001243428.1 | NP_001230357.1 | - | - | - | |
NM_001243429.1 | NP_001230358.1 | - | - | - | |
NM_001243432.2 | NP_001230361.1 | 4753309 | c.1201G>A | p.G401R | |
2845101 | c.1187A>C | p.K396T | |||
2157451 | c.1163T>C | p.V388A | |||
3713441 | c.1232C>T | p.P411L | |||
252495 | c.1238C>A | p.S413* | |||
6979599 | c.1212G>T | p.Q404H | |||
6917625 | c.1238C>T | p.S413L | |||
6942825 | c.1126C>T | p.L376F | |||
6906539 | c.1259C>G | p.S420* | |||
1483934 | c.1267C>A | p.P423T | |||
4918247 | c.1275G>A | p.M425I | |||
4385203 | c.1207G>A | p.A403T | |||
6947987 | c.1130G>C | p.R377P | |||
6912149 | c.1214C>T | p.A405V | |||
2845104 | c.1122C>T | p.R374R | |||
1414058 | c.1232delC | p.P411fs*25 | |||
6962461 | c.1271A>T | p.Y424F | |||
6656069 | c.1123G>A | p.A375T | |||
237354 | c.1266C>T | p.L422L | |||
5542895 | c.1203G>A | p.G401G | |||
6930369 | c.1192G>C | p.D398H | |||
3721669 | c.1179C>T | p.Y393Y | |||
4595830 | c.1229C>T | p.P410L | |||
3550664 | c.1225C>T | p.H409Y | |||
1030632 | c.1192G>A | p.D398N | |||
6981083 | c.1267_1268CC>TT | p.P423L | |||
6953565 | c.1255_1256CC>TT | p.P419F | |||
1616056 | c.1174C>T | p.R392C | |||
1414059 | c.1216C>A | p.L406I | |||
6964239 | c.1180G>A | p.A394T | |||
2845099 | c.1256C>T | p.P419L | |||
5594544 | c.1285C>T | p.H429Y | |||
4619133 | c.1233G>A | p.P411P | |||
1718764 | c.1254C>T | p.Y418Y | |||
6950977 | c.1175G>A | p.R392H | |||
6912187 | c.1159A>G | p.K387E | |||
3550665 | c.1215C>T | p.A405A | |||
4101565 | c.1287C>T | p.H429H | |||
6947253 | c.1199A>G | p.H400R | |||
419233 | c.1153A>G | p.M385V | |||
1414061 | c.1129C>T | p.R377C | |||
6964716 | c.1226A>G | p.H409R | |||
3799900 | c.1201G>T | p.G401W | |||
2845100 | c.1205T>A | p.I402N | |||
6559125 | c.1125C>A | p.A375A | |||
6094654 | c.1268C>G | p.P423R | |||
244305 | c.1223C>A | p.P408H | |||
6913063 | c.1129C>G | p.R377G | |||
5909847 | c.1231C>T | p.P411S | |||
4101566 | c.1137C>T | p.Y379Y | |||
6978057 | c.1280C>T | p.S427F | |||
419234 | c.1259C>T | p.S420L | |||
NM_004449.4 | NP_004440.1 | 48371 | c.1058G>T | p.R353L | |
NM_182918.3 | NP_891548.1 | 6917626 | c.1217C>T | p.S406L | |
5542896 | c.1182G>A | p.G394G | |||
4891794 | c.1204C>T | p.H402Y | |||
6912150 | c.1193C>T | p.A398V | |||
4753310 | c.1180G>A | p.G394R | |||
4296143 | c.1171G>A | p.D391N | |||
4894607 | c.1194C>T | p.A398A | |||
4781369 | c.1153C>T | p.R385C | |||
6978058 | c.1259C>T | p.S420F | |||
4619134 | c.1212G>A | p.P404P | |||
4918248 | c.1254G>A | p.M418I | |||
4812216 | c.1180G>T | p.G394W | |||
5594545 | c.1264C>T | p.H422Y | |||
6559126 | c.1104C>A | p.A368A | |||
6094655 | c.1247C>G | p.P416R | |||
4336874 | c.1211delC | p.P404fs*25 | |||
5909848 | c.1210C>T | p.P404S | |||
4296144 | c.1166A>C | p.K389T | |||
6942826 | c.1105C>T | p.L369F | |||
6979600 | c.1191G>T | p.Q397H | |||
6947988 | c.1109G>C | p.R370P | |||
4816081 | c.1246C>A | p.P416T | |||
4907109 | c.1116C>T | p.Y372Y | |||
6962462 | c.1250A>T | p.Y417F | |||
4812242 | c.1238C>T | p.S413L | |||
6930370 | c.1171G>C | p.D391H | |||
6953566 | c.1234_1235CC>TT | p.P412F | |||
4781843 | c.1142T>C | p.V381A | |||
4385204 | c.1186G>A | p.A396T | |||
4782807 | c.1195C>A | p.L399I | |||
4296147 | c.1101C>T | p.R367R | |||
6981084 | c.1246_1247CC>TT | p.P416L | |||
4892325 | c.1108C>T | p.R370C | |||
6912188 | c.1138A>G | p.K380E | |||
6656070 | c.1102G>A | p.A368T | |||
4812098 | c.1132A>G | p.M378V | |||
6964240 | c.1159G>A | p.A387T | |||
4881185 | c.1211C>T | p.P404L | |||
6947254 | c.1178A>G | p.H393R | |||
4296142 | c.1184T>A | p.I395N | |||
6950978 | c.1154G>A | p.R385H | |||
6913064 | c.1108C>G | p.R370G | |||
4595831 | c.1208C>T | p.P403L | |||
6906540 | c.1238C>G | p.S413* | |||
4907928 | c.1266C>T | p.H422H | |||
6964717 | c.1205A>G | p.H402R | |||
XM_011529486.2 | XP_011527788.1 | - | - | - | |
XM_017028287.1 | XP_016883776.1 | - | - | - | |
XM_017028288.1 | XP_016883777.1 | - | - | - | |
XM_017028289.1 | XP_016883778.1 | - | - | - | |
GenBank | AK297807.1 | - | - | - | |
AK300395.1 | - | - | - | ||
AK301277.1 | - | - | - | ||
AK303518.1 | - | - | - | ||
AK309469.1 | - | - | - | ||
AY204741.1 | AAP41719.1 | - | - | - | |
BC040168.1 | AAH40168.1 | - | - | - | |
ENST00000442448.0 | - | - | - | ||
KP100545.1 | - | - | - |
Literature Links: |
ERG PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Less than 200 bp |