Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
EWSR1 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001163285.1 | NP_001156757.1 | - | - | - |
NM_001163286.1 | NP_001156758.1 | - | - | - | |
NM_001163287.1 | NP_001156759.1 | 5776944 | c.897C>T | p.N299N | |
3842410 | c.962G>T | p.R321L | |||
5718486 | c.926G>A | p.R309H | |||
3553119 | c.857C>T | p.S286F | |||
7305530 | c.951G>C | p.R317R | |||
4298353 | c.970A>C | p.M324L | |||
5859124 | c.802C>T | p.R268* | |||
1251578 | c.835G>A | p.G279R | |||
6328329 | c.911G>A | p.R304K | |||
4800003 | c.924T>A | p.D308E | |||
1415484 | c.852A>G | p.G284G | |||
1592924 | c.799T>A | p.F267I | |||
5029932 | c.851G>C | p.G284A | |||
7075881 | c.892G>C | p.D298H | |||
6867021 | c.807G>T | p.Q269H | |||
3553116 | c.797C>T | p.S266L | |||
5619894 | c.913G>C | p.G305R | |||
2936267 | c.880A>T | p.M294L | |||
5729572 | c.898C>T | p.R300W | |||
4445903 | c.794-3C>G | p.? | |||
1205785 | c.874C>T | p.R292W | |||
6293143 | c.964G>A | p.G322S | |||
6432556 | c.899G>C | p.R300P | |||
1415486 | c.925C>T | p.R309C | |||
5065049 | c.963C>T | p.R321R | |||
NM_005243.3 | NP_005234.1 | 1415485 | c.925C>T | p.R309C | |
6432555 | c.899G>C | p.R300P | |||
3553115 | c.797C>T | p.S266L | |||
5029931 | c.851G>C | p.G284A | |||
370080 | c.803G>T | p.R268L | |||
6867020 | c.807G>T | p.Q269H | |||
7075880 | c.892G>C | p.D298H | |||
3842409 | c.962G>T | p.R321L | |||
224095 | c.892G>A | p.D298N | |||
6293142 | c.964G>A | p.G322S | |||
2936266 | c.880A>T | p.M294L | |||
1205784 | c.874C>T | p.R292W | |||
5619893 | c.913G>C | p.G305R | |||
5729571 | c.898C>T | p.R300W | |||
1251577 | c.835G>A | p.G279R | |||
5859123 | c.802C>T | p.R268* | |||
1415483 | c.852A>G | p.G284G | |||
4298352 | c.970A>C | p.M324L | |||
3553118 | c.857C>T | p.S286F | |||
4445902 | c.794-3C>G | p.? | |||
5065048 | c.963C>T | p.R321R | |||
1033129 | c.799T>A | p.F267I | |||
4800002 | c.924T>A | p.D308E | |||
7305529 | c.951G>C | p.R317R | |||
6328328 | c.911G>A | p.R304K | |||
5776943 | c.897C>T | p.N299N | |||
5718485 | c.926G>A | p.R309H | |||
NM_013986.3 | NP_053733.2 | 3553117 | c.875C>T | p.S292F | |
5776942 | c.915C>T | p.N305N | |||
5859122 | c.820C>T | p.R274* | |||
6432554 | c.917G>C | p.R306P | |||
4800001 | c.942T>A | p.D314E | |||
3842408 | c.980G>T | p.R327L | |||
6867019 | c.825G>T | p.Q275H | |||
2936265 | c.898A>T | p.M300L | |||
4298351 | c.988A>C | p.M330L | |||
4445901 | c.812-3C>G | p.? | |||
7305528 | c.969G>C | p.R323R | |||
5029930 | c.869G>C | p.G290A | |||
3553114 | c.815C>T | p.S272L | |||
5729570 | c.916C>T | p.R306W | |||
5157237 | c.870A>G | p.G290G | |||
1592923 | c.817T>A | p.F273I | |||
1616332 | c.892C>T | p.R298W | |||
7075879 | c.910G>C | p.D304H | |||
6293141 | c.982G>A | p.G328S | |||
5619892 | c.931G>C | p.G311R | |||
2936268 | c.943C>T | p.R315C | |||
5718484 | c.944G>A | p.R315H | |||
5065047 | c.981C>T | p.R327R | |||
6328327 | c.929G>A | p.R310K | |||
XM_005261389.3 | XP_005261446.1 | - | - | - | |
XM_005261390.4 | XP_005261447.1 | - | - | - | |
XM_011529995.2 | XP_011528297.1 | - | - | - | |
XM_011529996.2 | XP_011528298.1 | - | - | - | |
XM_011529997.1 | XP_011528299.1 | - | - | - | |
XM_011529998.1 | XP_011528300.1 | - | - | - | |
XM_011529999.2 | XP_011528301.1 | - | - | - | |
XM_011530000.1 | XP_011528302.1 | - | - | - | |
XM_011530001.2 | XP_011528303.1 | - | - | - | |
XM_011530002.2 | XP_011528304.1 | - | - | - | |
XM_017028644.1 | XP_016884133.1 | - | - | - | |
XM_017028645.1 | XP_016884134.1 | - | - | - | |
XM_017028646.1 | XP_016884135.1 | - | - | - | |
XM_017028647.1 | XP_016884136.1 | - | - | - | |
XM_017028648.1 | XP_016884137.1 | - | - | - | |
XM_017028649.1 | XP_016884138.1 | - | - | - | |
XM_017028650.1 | XP_016884139.1 | - | - | - | |
XM_017028651.1 | XP_016884140.1 | - | - | - | |
XM_017028652.1 | XP_016884141.1 | - | - | - | |
XM_017028653.1 | XP_016884142.1 | - | - | - | |
XM_017028654.1 | XP_016884143.1 | - | - | - | |
XM_017028655.1 | XP_016884144.1 | - | - | - | |
XM_017028656.1 | XP_016884145.1 | - | - | - | |
XM_017028657.1 | XP_016884146.1 | - | - | - | |
XM_017028658.1 | XP_016884147.1 | - | - | - | |
XM_017028659.1 | XP_016884148.1 | - | - | - | |
XM_017028660.1 | XP_016884149.1 | - | - | - | |
XM_017028661.1 | XP_016884150.1 | - | - | - | |
XM_017028662.1 | XP_016884151.1 | - | - | - | |
XM_017028663.1 | XP_016884152.1 | - | - | - | |
XM_017028664.1 | XP_016884153.1 | - | - | - | |
XM_017028665.1 | XP_016884154.1 | - | - | - | |
XM_017028666.1 | XP_016884155.1 | - | - | - | |
GenBank | AK056309.1 | BAB71145.1 | - | - | - |
AK056681.1 | BAB71252.1 | - | - | - | |
AK127624.1 | - | - | - | ||
AK225096.1 | - | - | - | ||
AK225102.1 | - | - | - | ||
AL833489.1 | - | - | - | ||
BC000527.2 | AAH00527.1 | - | - | - | |
BC004817.1 | AAH04817.1 | - | - | - | |
BC011048.1 | AAH11048.1 | - | - | - | |
BC072442.1 | AAH72442.1 | - | - | - | |
BX648769.1 | - | - | - | ||
CR456490.1 | CAG30376.1 | - | - | - | |
ENST00000397938.0 | - | - | - | ||
JX977847.1 | - | - | - | ||
X66899.1 | CAA47350.1 | - | - | - |
Literature Links: |
EWSR1 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |