Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CLASP2 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001207044.1 | NP_001193973.1 | 4866749 | c.3461C>A | p.P1154H |
7243664 | c.3343-1G>T | p.? | |||
5517318 | c.3379G>C | p.E1127Q | |||
7361402 | c.3351C>G | p.I1117M | |||
6097476 | c.3377G>A | p.R1126K | |||
NM_015097.2 | NP_055912.2 | - | - | - | |
XM_006713040.1 | XP_006713103.1 | - | - | - | |
XM_006713041.1 | XP_006713104.1 | - | - | - | |
XM_006713042.1 | XP_006713105.1 | - | - | - | |
XM_006713048.1 | XP_006713111.1 | - | - | - | |
XM_006713049.1 | XP_006713112.1 | - | - | - | |
XM_006713050.1 | XP_006713113.1 | - | - | - | |
XM_006713052.1 | XP_006713115.1 | - | - | - | |
XM_006713053.1 | XP_006713116.1 | - | - | - | |
XM_011533515.1 | XP_011531817.1 | - | - | - | |
XM_017005947.1 | XP_016861436.1 | - | - | - | |
XM_017005948.1 | XP_016861437.1 | - | - | - | |
XM_017005949.1 | XP_016861438.1 | - | - | - | |
XM_017005950.1 | XP_016861439.1 | - | - | - | |
XM_017005951.1 | XP_016861440.1 | - | - | - | |
XM_017005952.1 | XP_016861441.1 | - | - | - | |
XM_017005953.1 | XP_016861442.1 | - | - | - | |
XM_017005954.1 | XP_016861443.1 | - | - | - | |
XM_017005955.1 | XP_016861444.1 | - | - | - | |
XM_017005956.1 | XP_016861445.1 | - | - | - | |
XM_017005957.1 | XP_016861446.1 | - | - | - | |
XM_017005958.1 | XP_016861447.1 | - | - | - | |
XM_017005959.1 | XP_016861448.1 | 7361400 | c.4011C>G | p.I1337M | |
6097474 | c.4037G>A | p.R1346K | |||
7243662 | c.4003-1G>T | p.? | |||
XM_017005960.1 | XP_016861449.1 | - | - | - | |
XM_017005961.1 | XP_016861450.1 | - | - | - | |
XM_017005962.1 | XP_016861451.1 | - | - | - | |
XM_017005963.1 | XP_016861452.1 | - | - | - | |
XM_017005964.1 | XP_016861453.1 | - | - | - | |
XM_017005965.1 | XP_016861454.1 | - | - | - | |
XM_017005966.1 | XP_016861455.1 | - | - | - | |
XM_017005967.1 | XP_016861456.1 | - | - | - | |
XM_017005968.1 | XP_016861457.1 | - | - | - | |
XM_017005969.1 | XP_016861458.1 | - | - | - | |
XM_017005970.1 | XP_016861459.1 | - | - | - | |
XM_017005971.1 | XP_016861460.1 | - | - | - | |
XM_017005972.1 | XP_016861461.1 | - | - | - | |
XM_017005973.1 | XP_016861462.1 | - | - | - | |
XM_017005974.1 | XP_016861463.1 | - | - | - | |
XM_017005975.1 | XP_016861464.1 | - | - | - | |
XM_017005976.1 | XP_016861465.1 | - | - | - | |
XM_017005977.1 | XP_016861466.1 | - | - | - | |
XM_017005978.1 | XP_016861467.1 | - | - | - | |
XM_017005979.1 | XP_016861468.1 | - | - | - | |
XM_017005980.1 | XP_016861469.1 | - | - | - | |
XM_017005981.1 | XP_016861470.1 | - | - | - | |
XM_017005982.1 | XP_016861471.1 | - | - | - | |
XM_017005983.1 | XP_016861472.1 | - | - | - | |
XM_017005984.1 | XP_016861473.1 | - | - | - | |
XM_017005985.1 | XP_016861474.1 | - | - | - | |
XM_017005986.1 | XP_016861475.1 | - | - | - | |
GenBank | AB014527.2 | - | - | - | |
AK024770.1 | BAB14995.1 | - | - | - | |
AK293963.1 | - | - | - | ||
AL137636.1 | CAB70852.1 | - | - | - | |
BC011530.1 | AAH11530.1 | - | - | - | |
BC140778.1 | - | - | - | ||
BX640725.1 | CAE45842.1 | - | - | - |
Literature Links: |
CLASP2 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |