Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GALNT13 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001301627.1 | NP_001288556.1 | 3425344 | c.1132G>T | p.D378Y |
6316189 | c.1156+1G>T | p.? | |||
3961319 | c.1156+9T>A | p.? | |||
3568085 | c.1060C>T | p.P354S | |||
3425340 | c.1036C>T | p.R346W | |||
6192449 | c.1129A>C | p.K377Q | |||
6669007 | c.1137C>A | p.F379L | |||
4809199 | c.1140C>T | p.F380F | |||
3299360 | c.1119G>C | p.M373I | |||
3299372 | c.1151C>A | p.S384Y | |||
3694920 | c.1056T>G | p.T352T | |||
4607240 | c.1046C>A | p.T349N | |||
5662800 | c.1105G>T | p.A369S | |||
3425342 | c.1046C>T | p.T349I | |||
7244712 | c.1132G>C | p.D378H | |||
6087704 | c.1155A>C | p.P385P | |||
4900073 | c.1049C>T | p.P350L | |||
3938560 | c.1124A>T | p.E375V | |||
6572751 | c.1098G>A | p.R366R | |||
6669009 | c.1059T>C | p.F353F | |||
3299362 | c.1123G>A | p.E375K | |||
3299376 | c.1155A>G | p.P385P | |||
4304591 | c.1151C>T | p.S384F | |||
3299350 | c.1089G>T | p.K363N | |||
4085728 | c.1110A>C | p.E370D | |||
3729206 | c.1105G>A | p.A369T | |||
NM_052917.3 | NP_443149.2 | 4900072 | c.1049C>T | p.P350L | |
6192448 | c.1129A>C | p.K377Q | |||
716278 | c.1072G>T | p.G358C | |||
4809198 | c.1140C>T | p.F380F | |||
366336 | c.1120G>T | p.D374Y | |||
1400221 | c.1036C>T | p.R346W | |||
1007941 | c.1113T>G | p.V371V | |||
3938559 | c.1124A>T | p.E375V | |||
6087703 | c.1155A>C | p.P385P | |||
3425343 | c.1132G>T | p.D378Y | |||
4304590 | c.1151C>T | p.S384F | |||
6669006 | c.1137C>A | p.F379L | |||
4085727 | c.1110A>C | p.E370D | |||
1196877 | c.1089G>T | p.K363N | |||
3299375 | c.1155A>G | p.P385P | |||
4607239 | c.1046C>A | p.T349N | |||
3299361 | c.1123G>A | p.E375K | |||
3729205 | c.1105G>A | p.A369T | |||
716277 | c.1132G>C | p.D378H | |||
5662799 | c.1105G>T | p.A369S | |||
3694919 | c.1056T>G | p.T352T | |||
3961318 | c.1156+9T>A | p.? | |||
1007940 | c.1078G>A | p.V360I | |||
3425341 | c.1046C>T | p.T349I | |||
3568084 | c.1060C>T | p.P354S | |||
6669008 | c.1059T>C | p.F353F | |||
1253040 | c.1073G>C | p.G358A | |||
6572750 | c.1098G>A | p.R366R | |||
1163609 | c.1074T>C | p.G358G | |||
3299371 | c.1151C>A | p.S384Y | |||
3299359 | c.1119G>C | p.M373I | |||
6316188 | c.1156+1G>T | p.? | |||
XM_011510537.2 | XP_011508839.1 | - | - | - | |
XM_011510538.2 | XP_011508840.1 | - | - | - | |
XM_017003258.1 | XP_016858747.1 | - | - | - | |
XM_017003259.1 | XP_016858748.1 | - | - | - | |
XM_017003260.1 | XP_016858749.1 | - | - | - | |
XM_017003261.1 | XP_016858750.1 | - | - | - | |
XM_017003262.1 | XP_016858751.1 | - | - | - | |
XM_017003263.1 | XP_016858752.1 | - | - | - | |
GenBank | AB067505.1 | BAB67811.1 | - | - | - |
AB078142.1 | BAC54545.1 | - | - | - | |
AJ505991.2 | CAD44533.2 | - | - | - | |
AK131195.1 | - | - | - | ||
AK308675.1 | - | - | - | ||
BC101031.1 | AAI01032.1 | - | - | - | |
BC101032.2 | - | - | - | ||
BC101033.1 | AAI01034.1 | - | - | - | |
BC101034.2 | - | - | - | ||
ENST00000392825.0 | - | - | - | ||
KJ534843.1 | - | - | - | ||
KJ535058.1 | - | - | - |
Literature Links: |
GALNT13 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |