Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ecd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027475.3 | NP_081751.1 | 2, 3 | 547 | ||
GenBank mRNA | AK028664.1 | - | 2, 3 | 511 | ||
AK029169.1 | - | 2, 3 | 479 | |||
AK043261.1 | - | 2, 3 | 396 | |||
AK213825.1 | - | 2, 3 | 242 | |||
BC013470.1 | - | 2, 3 | 496 | 4220504 |