Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CYBA SIRNA Assays ›
          Gene Symbol:
          CYBA
          Gene Name:
          cytochrome b-245 alpha chain
          Gene Aliases:
          p22-PHOX
          Chromosome Location:
          Chr.16: 88643289 - 88651084 on Build GRCh38
          UniGene ID:
          Hs.513803
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 105948

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CYBA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000101.3 NP_000092.2 4 333
          XM_011522905.2 XP_011521207.1 4 328
          GenBank mRNA AK097127.1 - 4 236
          AK300049.1 - 4 237
          AK309030.1 - 4 237
          BC006465.1 - 4 289 3502788
          BT006861.1 - 4 262
          BX109994.1 - 4 298
          M21186.1 - 4 290

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          positive regulation of endothelial cell proliferation
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
          negative regulation of glomerular filtration by angiotensin
          superoxide metabolic process
          inflammatory response
          smooth muscle hypertrophy
          cytochrome complex assembly
          positive regulation of cell growth
          response to nutrient levels
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          positive regulation of superoxide anion generation
          positive regulation of toll-like receptor 2 signaling pathway
          response to drug
          superoxide anion generation
          innate immune response
          cell redox homeostasis
          respiratory burst
          vascular endothelial growth factor receptor signaling pathway
          positive regulation of smooth muscle cell proliferation
          hydrogen peroxide biosynthetic process
          positive regulation of phagocytosis
          regulation of release of sequestered calcium ion into cytosol
          oxidation-reduction process
          positive regulation of mucus secretion
          response to interleukin-1
          cellular response to amino acid stimulus
          cellular response to mechanical stimulus
          cellular response to glucose stimulus
          cellular response to tumor necrosis factor
          cellular response to organic cyclic compound
          cellular response to gamma radiation
          positive regulation of defense response to bacterium
          positive regulation of reactive oxygen species biosynthetic process
          protein binding
          electron carrier activity
          superoxide-generating NADPH oxidase activity
          SH3 domain binding
          heme binding
          metal ion binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0