Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LRRFIP2 SIRNA Assays ›
Gene Symbol:
LRRFIP2
Gene Name:
LRR binding FLII interacting protein 2
Gene Aliases:
HUFI-2
Chromosome Location:
Chr.3: 37052626 - 37176817 on Build GRCh38
UniGene ID:
Hs.740522
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
108004
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LRRFIP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134369.2
NP_001127841.1
14
1505
NM_001282691.1
NP_001269620.1
18
1494
NM_006309.3
NP_006300.1
27
2095
NM_017724.2
NP_060194.1
13
1433
XM_005265539.2
XP_005265596.1
28
3095
XM_005265540.1
XP_005265597.1
29
2378
XM_005265550.1
XP_005265607.1
20
1931
XM_005265551.1
XP_005265608.1
19
1820
XM_005265553.1
XP_005265610.1
16
1667
XM_005265554.2
XP_005265611.1
16
2399
XM_005265556.1
XP_005265613.1
15
1595
XM_006713385.1
XP_006713448.1
28
2360
XM_006713386.1
XP_006713449.1
27
2321
XM_006713387.1
XP_006713450.1
27
2318
XM_006713388.1
XP_006713451.1
27
2315
XM_006713389.1
XP_006713452.1
27
2312
XM_006713390.1
XP_006713453.1
28
2355
XM_006713392.1
XP_006713455.1
27
2291
XM_006713393.1
XP_006713456.1
27
2273
XM_006713394.1
XP_006713457.1
26
2270
XM_006713395.1
XP_006713458.1
27
2270
XM_006713396.1
XP_006713459.1
26
2255
XM_006713397.1
XP_006713460.1
25
2147
XM_006713400.1
XP_006713463.1
17
1712
XM_006713404.1
XP_006713467.1
14
1445
XM_011534217.1
XP_011532519.1
28
2363
XM_011534218.1
XP_011532520.1
29
2406
XM_011534219.1
XP_011532521.1
29
2474
XM_011534221.1
XP_011532523.1
26
2255
XM_011534222.1
XP_011532524.1
26
2168
XM_011534229.1
XP_011532531.1
18
1755
XM_017007465.1
XP_016862954.1
25
2210
XM_017007466.1
XP_016862955.1
26
2204
XM_017007467.1
XP_016862956.1
26
2198
XM_017007468.1
XP_016862957.1
23
2102
XM_017007469.1
XP_016862958.1
25
2096
XM_017007470.1
XP_016862959.1
23
2060
XM_017007471.1
XP_016862960.1
18
1769
XM_017007472.1
XP_016862961.1
18
1727
XM_017007473.1
XP_016862962.1
17
1697
XM_017007474.1
XP_016862963.1
17
1676
XM_017007475.1
XP_016862964.1
17
2408
XM_017007476.1
XP_016862965.1
17
1710
XM_017007477.1
XP_016862966.1
16
2336
XM_017007478.1
XP_016862967.1
15
1574
XM_017007479.1
XP_016862968.1
16
1574
XM_017007480.1
XP_016862969.1
15
1818
XM_017007481.1
XP_016862970.1
15
1565
XM_017007482.1
XP_016862971.1
14
1502
XM_017007483.1
XP_016862972.1
14
2204
XM_017007484.1
XP_016862973.1
15
1515
GenBank mRNA
AF115509.1
-
27
2054
AK000255.1
-
12
1118
AK024692.1
-
6
451
AK124432.1
-
5
3229
AK291514.1
-
14
1482
AK302282.1
-
18
1468
BC053668.1
-
13
1390
6146978
BX648794.1
-
10
884
CR749705.1
-
4
1822
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
biological_process
Wnt signaling pathway
protein binding
LRR domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-qlhls:80/10.239.162.255:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0