Search
Search

SNRK| Gene Symbol: SNRK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100594.1 | NP_001094064.1 | 2 | 777 | ||
| NM_017719.4 | NP_060189.3 | 3 | 839 | |||
| XM_005265245.3 | XP_005265302.1 | 4 | 889 | |||
| GenBank mRNA | AK000231.1 | - | 1 | 599 | ||
| AK291360.1 | - | 4 | 890 | |||
| AK314376.1 | - | 2 | 662 | |||
| BC071567.1 | - | 2 | 751 | 4375623 | ||
| CR749621.1 | - | 2 | 699 | |||
| D43636.2 | - | 1 | 511 | |||