Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286752.1 | NP_001273681.1 | 12 | 1886 | ||
NM_017438.4 | NP_059134.1 | 11 | 1743 | |||
NR_040087.2 | 10 | 1654 | ||||
XM_011529636.1 | XP_011527938.1 | 11 | 1792 | |||
XM_011529637.1 | XP_011527939.1 | 11 | 1857 | |||
XM_011529638.2 | XP_011527940.1 | 11 | 1765 | |||
XM_011529639.1 | XP_011527941.1 | 11 | 1543 | |||
XM_011529640.2 | XP_011527942.1 | 11 | 1808 | |||
XM_011529642.1 | XP_011527944.1 | 12 | 1886 | |||
XM_011529643.1 | XP_011527945.1 | 12 | 1837 | |||
XM_011529644.1 | XP_011527946.1 | 12 | 1398 | |||
XM_017028403.1 | XP_016883892.1 | 11 | 1334 | |||
XM_017028404.1 | XP_016883893.1 | 12 | 1442 | |||
GenBank mRNA | AF391112.1 | - | 10 | 1221 | ||
AK001660.1 | - | 11 | 1310 | |||
AK095215.1 | - | 10 | 2610 | |||
AK124254.1 | - | 1 | 613 | |||
AK126368.1 | - | 4 | 1820 | |||
BC036556.2 | - | 12 | 1560 | 5266627 |