Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318046.1 | NP_001304975.1 | 5 | 452 | ||
NM_001318047.1 | NP_001304976.1 | 5 | 455 | |||
NM_001318048.1 | NP_001304977.1 | 5 | 452 | |||
NM_007356.2 | NP_031382.2 | 5 | 455 | |||
XM_011515975.1 | XP_011514277.1 | 5 | 455 | |||
XM_011515978.1 | XP_011514280.1 | 5 | 455 | |||
XM_011515979.2 | XP_011514281.1 | 5 | 455 | |||
XM_011515980.2 | XP_011514282.1 | 5 | 455 | |||
XM_017011879.1 | XP_016867368.1 | 5 | 455 | |||
XM_017011880.1 | XP_016867369.1 | 5 | 455 | |||
GenBank mRNA | AF028816.1 | - | 5 | 451 | ||
AK300385.1 | - | 5 | 456 | |||
BC140804.1 | - | 5 | 442 | 9021672 | ||
BC142619.1 | - | 5 | 437 | |||
BC144591.1 | - | 5 | 442 | 9053121 | ||
DA859196.1 | - | 5 | 456 |