Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other JAG1 SIRNA Assays ›
Gene Symbol:
Jag1
Gene Name:
jagged 1
Gene Aliases:
ABE2, Gsfabe2, Htu, Ozz, Ser-1
Chromosome Location:
Chr.2: 137081451 - 137116520 on Build GRCm38
UniGene ID:
Mm.22398
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
156900
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Jag1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_013822.5
NP_038850.1
4
746
GenBank mRNA
AF171092.1
-
4
751
AK137092.1
-
4
646
BC058675.1
-
4
558
6834418
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel remodeling
morphogenesis of an epithelial sheet
T cell mediated immunity
pulmonary valve morphogenesis
cardiac right ventricle morphogenesis
cell communication
Notch signaling pathway
multicellular organism development
organ morphogenesis
response to muramyl dipeptide
aorta morphogenesis
regulation of cell proliferation
auditory receptor cell differentiation
camera-type eye development
negative regulation of cell differentiation
negative regulation of fat cell differentiation
positive regulation of myeloid cell differentiation
negative regulation of neuron differentiation
positive regulation of osteoblast differentiation
positive regulation of Notch signaling pathway
positive regulation of transcription from RNA polymerase II promoter
inner ear development
cardiac septum morphogenesis
ciliary body morphogenesis
pulmonary artery morphogenesis
cardiac neural crest cell development involved in outflow tract morphogenesis
Notch signaling involved in heart development
endocardial cushion cell development
negative regulation of stem cell differentiation
Notch binding
calcium ion binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0