Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HSPA8 SIRNA Assays ›
          Gene Symbol:
          Hspa8
          Gene Name:
          heat shock protein 8
          Gene Aliases:
          2410008N15Rik, Hsc70, Hsc71, Hsc73, Hsp73, Hspa10
          Chromosome Location:
          Chr.9: 40801273 - 40805199 on Build GRCm38
          UniGene ID:
          Mm.290774
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 159533

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hspa8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031165.4 NP_112442.2 9 1901
          GenBank mRNA AK004608.1 - 4 2957
          AK035286.1 - 9 1899
          AK075935.1 - 9 1898
          AK081701.1 - 9 1900
          AK098074.1 - 9 1898
          AK135281.1 - 5 1070
          AK145416.1 - 9 1821
          AK145579.1 - 9 1895
          AK146547.1 - 9 1905
          AK146708.1 - 9 1896
          AK146985.1 - 9 1899
          AK147864.1 - 9 1899
          AK150474.1 - 9 1893
          AK150498.1 - 9 1893
          AK150701.1 - 9 1903
          AK150846.1 - 4 635
          AK150958.1 - 9 1896
          AK151041.1 - 9 1899
          AK151065.1 - 9 1899
          AK151127.1 - 9 1902
          AK151287.1 - 9 1899
          AK151435.1 - 9 1899
          AK151516.1 - 9 1905
          AK151537.1 - 9 1896
          AK151741.1 - 9 1899
          AK151775.1 - 9 1901
          AK151808.1 - 9 1902
          AK151865.1 - 9 1899
          AK151892.1 - 9 1901
          AK151948.1 - 9 1902
          AK151997.1 - 9 1899
          AK152315.1 - 9 1902
          AK152598.1 - 9 1899
          AK152697.1 - 9 1899
          AK152703.1 - 9 1899
          AK152768.1 - 9 1902
          AK152803.1 - 9 1905
          AK153032.1 - 9 1901
          AK153834.1 - 9 1899
          AK157831.1 - 9 1899
          AK159479.1 - 9 1899
          AK162775.1 - 9 1899
          AK163987.1 - 9 1899
          AK164000.1 - 9 1899
          AK166545.1 - 9 1901
          AK166643.1 - 9 1913
          AK166721.1 - 9 1896
          AK166767.1 - 9 1899
          AK166776.1 - 9 1902
          AK166808.1 - 9 1899
          AK166830.1 - 9 1897
          AK166846.1 - 9 1899
          AK166861.1 - 9 1902
          AK166873.1 - 9 1899
          AK166908.1 - 9 1902
          AK166910.1 - 9 1899
          AK166913.1 - 9 1899
          AK166933.1 - 9 1899
          AK167043.1 - 9 1901
          AK167121.1 - 9 1902
          AK167122.1 - 9 1899
          AK167134.1 - 9 1897
          AK167163.1 - 9 1901
          AK167218.1 - 9 1899
          AK167229.1 - 9 1900
          AK167254.1 - 9 1900
          AK167845.1 - 9 1902
          AK167910.1 - 9 1902
          AK168435.1 - 9 1902
          AK168492.1 - 9 1901
          AK168511.1 - 7 1437
          AK168519.1 - 9 1899
          AK168542.1 - 9 1901
          AK168577.1 - 9 1900
          AK168625.1 - 9 1900
          AK168654.1 - 9 1900
          AK168711.1 - 9 1899
          AK168750.1 - 9 1902
          AK168776.1 - 9 1902
          AK168887.1 - 9 1901
          AK168934.1 - 9 1902
          AK169093.1 - 9 1899
          AK169179.1 - 9 1899
          AK169236.1 - 9 1899
          AK169293.1 - 9 1913
          AK169329.1 - 9 1902
          AK169594.1 - 9 1898
          AK184860.1 - 2 190
          AK190678.1 - 2 296
          AK190709.1 - 2 213
          AK192102.1 - 2 198
          AK192871.1 - 2 173
          AK193113.1 - 2 296
          AK194470.1 - 2 168
          AK194530.1 - 2 158
          AK195783.1 - 2 177
          AK197043.1 - 2 158
          AK197541.1 - 2 296
          AK197732.1 - 2 296
          AK198838.1 - 2 296
          AK199036.1 - 2 167
          AK200192.1 - 2 171
          AK200291.1 - 2 184
          AK201037.1 - 2 175
          AK201282.1 - 2 156
          AK201788.1 - 2 179
          AK202524.1 - 2 180
          AK204394.1 - 2 297
          AK209652.1 - 2 177
          AK217969.1 - 2 186
          AK218570.1 - 2 296
          AK218761.1 - 2 192
          AK219581.1 - 2 180
          BC006722.1 - 9 1923 3711399
          BC066191.1 - 9 1855 30467052
          BC085486.1 - 9 1887 30540914
          BC089322.1 - 9 1880 30538640
          BC089457.1 - 9 1858 30628505
          BC094900.1 - 9 1815 30063483
          BC106169.1 - 7 1579
          BC106193.1 - 9 1883 6329283
          U27129.1 - 9 1850

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mRNA processing
          protein folding
          RNA splicing
          protein refolding
          negative regulation of transcription, DNA-templated
          ATP metabolic process
          positive regulation of mRNA splicing, via spliceosome
          chaperone mediated protein folding requiring cofactor
          regulation of cell cycle
          chaperone-mediated protein folding
          clathrin coat disassembly
          negative regulation of fibril organization
          nucleotide binding
          G-protein coupled receptor binding
          receptor binding
          protein binding
          ATP binding
          ATPase activity
          enzyme binding
          MHC class II protein complex binding
          heat shock protein binding
          ubiquitin protein ligase binding
          ATPase activity, coupled
          ADP binding
          poly(A) RNA binding
          unfolded protein binding
          C3HC4-type RING finger domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0