Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other YAP1 SIRNA Assays ›
Gene Symbol:
YAP1
Gene Name:
Yes associated protein 1
Gene Aliases:
COB1, YAP, YAP2, YAP65, YKI
Chromosome Location:
Chr.11: 102109957 - 102233423 on Build GRCh38
UniGene ID:
Hs.503692
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
17470
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: YAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130145.2
NP_001123617.1
7
1467
NM_001195044.1
NP_001181973.1
6
1419
NM_001195045.1
NP_001181974.1
7
1064
NM_001282097.1
NP_001269026.1
6
1353
NM_001282098.1
NP_001269027.1
5
1317
NM_001282099.1
NP_001269028.1
6
1365
NM_001282100.1
NP_001269029.1
6
1431
NM_001282101.1
NP_001269030.1
7
1479
NM_006106.4
NP_006097.2
5
1305
XM_005271378.3
XP_005271435.1
7
1989
XM_005271380.3
XP_005271437.1
7
1977
XM_005271381.3
XP_005271438.1
6
1941
XM_005271383.3
XP_005271440.1
6
1929
XM_011542555.2
XP_011540857.1
7
1077
XM_011542556.2
XP_011540858.1
7
1071
XM_017017093.1
XP_016872582.1
6
1011
GenBank mRNA
AB567720.1
-
6
965
AB567721.1
-
7
1079
AK300414.1
-
7
782
AK304485.1
-
4
340
AK316116.1
-
7
990
AL832620.2
-
6
760
AY316529.1
-
6
1031
BC026212.2
-
1
48
BC038235.1
-
7
1372
5747370
DC419625.1
-
4
340
HE864159.1
-
3
314
HE864160.1
-
4
350
HE864161.1
-
4
362
HE864162.1
-
4
428
HE864163.1
-
5
476
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
vasculogenesis
embryonic heart tube morphogenesis
transcription initiation from RNA polymerase II promoter
cellular response to DNA damage stimulus
cell proliferation
positive regulation of cell proliferation
regulation of keratinocyte proliferation
keratinocyte differentiation
negative regulation of epithelial cell differentiation
notochord development
somatic stem cell population maintenance
hippo signaling
positive regulation of transcription from RNA polymerase II promoter
positive regulation of organ growth
paraxial mesoderm development
lateral mesoderm development
regulation of neurogenesis
contact inhibition
bud elongation involved in lung branching
lung epithelial cell differentiation
cellular response to retinoic acid
cellular response to gamma radiation
regulation of stem cell proliferation
regulation of metanephric nephron tubule epithelial cell differentiation
positive regulation of canonical Wnt signaling pathway
positive regulation of stem cell population maintenance
negative regulation of nucleic acid-templated transcription
negative regulation of stem cell differentiation
negative regulation of extrinsic apoptotic signaling pathway
RNA polymerase II core promoter sequence-specific DNA binding
transcription factor activity, RNA polymerase II transcription factor binding
chromatin binding
transcription coactivator activity
transcription corepressor activity
protein binding
protein C-terminus binding
transcription regulatory region DNA binding
proline-rich region binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0