Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spcs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025668.3 | NP_079944.1 | 5 | 1368 | ||
GenBank mRNA | AK017504.1 | - | 5 | 1373 | ||
AK077510.1 | - | 5 | 1378 | |||
AK134578.1 | - | 5 | 1369 | |||
AK168173.1 | - | 5 | 1376 | |||
AK180141.1 | - | 1 | 337 | |||
BC010547.1 | - | 4 | 1189 | |||
BC050150.1 | - | 5 | 1377 | 30026746 |