Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TLR2 SIRNA Assays ›
          Gene Symbol:
          Tlr2
          Gene Name:
          toll-like receptor 2
          Gene Aliases:
          Ly105
          Chromosome Location:
          Chr.3: 83836272 - 83841825 on Build GRCm38
          UniGene ID:
          Mm.87596
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 187991

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tlr2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011905.3 NP_036035.3 3 906
          GenBank mRNA AF124741.1 - 3 845
          AF165189.1 - 3 870
          AF185284.1 - 3 860
          AF216289.1 - 3 869
          AK005043.1 - 3 907
          AK154504.1 - 3 876
          AY179346.1 - 2 721
          BC014693.1 - 3 1065 3966249

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          microglial cell activation
          toll-like receptor signaling pathway
          response to molecule of bacterial origin
          response to molecule of fungal origin
          cytokine secretion involved in immune response
          immune system process
          positive regulation of leukocyte migration
          regulation of dendritic cell cytokine production
          cell surface pattern recognition receptor signaling pathway
          MyD88-dependent toll-like receptor signaling pathway
          leukotriene metabolic process
          inflammatory response
          immune response
          signal transduction
          I-kappaB phosphorylation
          negative regulation of cell proliferation
          response to bacterium
          positive regulation of Wnt signaling pathway
          central nervous system myelin formation
          response to bacterial lipoprotein
          response to peptidoglycan
          interleukin-10 production
          negative regulation of interleukin-12 production
          negative regulation of interleukin-17 production
          positive regulation of chemokine production
          positive regulation of interferon-beta production
          positive regulation of interleukin-10 production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-18 production
          positive regulation of interleukin-6 production
          positive regulation of interleukin-8 production
          positive regulation of tumor necrosis factor production
          positive regulation of toll-like receptor signaling pathway
          toll-like receptor 2 signaling pathway
          positive regulation of NF-kappaB import into nucleus
          detection of triacyl bacterial lipopeptide
          detection of diacyl bacterial lipopeptide
          positive regulation of tumor necrosis factor biosynthetic process
          chloramphenicol transport
          negative regulation of growth of symbiont in host
          innate immune response
          positive regulation of nitric oxide biosynthetic process
          positive regulation of transcription from RNA polymerase II promoter
          nitric oxide metabolic process
          positive regulation of oligodendrocyte differentiation
          regulation of cytokine secretion
          positive regulation of cytokine secretion
          positive regulation of inflammatory response
          defense response to Gram-positive bacterium
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of nitric-oxide synthase biosynthetic process
          induction by symbiont of defense-related host nitric oxide production
          positive regulation of macrophage cytokine production
          response to fatty acid
          cellular response to bacterial lipopeptide
          cellular response to lipoteichoic acid
          cellular response to peptidoglycan
          cellular response to diacyl bacterial lipopeptide
          cellular response to triacyl bacterial lipopeptide
          receptor activity
          transmembrane signaling receptor activity
          protein binding
          signaling pattern recognition receptor activity
          triacyl lipopeptide binding
          diacyl lipopeptide binding
          peptidoglycan binding
          protein heterodimerization activity
          lipoteichoic acid binding
          lipopeptide binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-hhcgv:80/100.66.79.225:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0