Search
Search

Usp18| Gene Symbol: Usp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011909.2 | NP_036039.2 | 11 | 1404 | ||
| GenBank mRNA | AF069502.1 | - | 11 | 1365 | ||
| AK004296.1 | - | 11 | 1370 | |||
| AK149772.1 | - | 11 | 1405 | |||
| AK151011.1 | - | 11 | 1405 | |||
| AK151260.1 | - | 11 | 1405 | |||
| AK151722.1 | - | 11 | 1405 | |||
| AK153312.1 | - | 11 | 1374 | |||
| BC138562.1 | - | 11 | 1286 | 8861582 | ||
| BC138577.1 | - | 11 | 1286 | 8861597 | ||