Search
Search

Pdgfrb| Gene Symbol: Pdgfrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031525.1 | NP_113713.1 | 3 | 713 | ||
| XM_006254789.2 | XP_006254851.1 | 3 | 803 | |||
| XM_008772084.1 | XP_008770306.1 | 2 | 354 | |||
| GenBank mRNA | AY090783.1 | - | 3 | 713 | ||
| Z14119.1 | - | 1 | 246 | |||