Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SDHA SIRNA Assays ›
          Gene Symbol:
          SDHA LOC220729 SDHAP1 SDHAP2
          Gene Name:
          succinate dehydrogenase complex flavoprotein subunit A
          succinate dehydrogenase complex flavoprotein subunit A pseudogene
          succinate dehydrogenase complex flavoprotein subunit A pseudogene 1
          succinate dehydrogenase complex flavoprotein subunit A pseudogene 2
          Gene Aliases:
          CMD1GG, FP, PGL5, SDH1, SDH2, SDHF
          -
          SDHAL1, SDHALP1
          SDHAL2, SDHALP2
          Chromosome Location:
          Chr.5: 218223 - 264816 on Build GRCh38
          Chromosome Location:
          Chr.3: 197614027 - 197627881 on Build GRCh38
          Chromosome Location:
          Chr.3: 195959921 - 195990279 on Build GRCh38
          Chromosome Location:
          Chr.3: 195658039 - 195688871 on Build GRCh38
          UniGene ID:
          Hs.440475
          Hs.478854
          Hs.566872
          Hs.596261
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID 212749

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SDHA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001294332.1 NP_001281261.1 2 266
          NM_004168.3 NP_004159.2 2 266
          XM_005248331.3 XP_005248388.1 2 266
          XM_011514072.1 XP_011512374.1 2 266
          XM_011514073.1 XP_011512375.1 2 266
          XM_017009685.1 XP_016865174.1 2 266
          GenBank mRNA AB208991.1 - 2 152
          AK225027.1 - 2 167
          AK291311.1 - 2 165
          AK295937.1 - 2 171
          AK302524.1 - 2 173
          BC001380.2 - 2 162 3051442
          BC041016.1 - 2 205 3138724
          D30648.1 - 2 139
          DA160919.1 - 2 266
          L21936.1 - 2 157
          Gene Symbol: LOC220729
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003266.2 2 171
          GenBank mRNA AK299863.1 - 2 171
          AK309924.1 - 2 161
          Gene Symbol: SDHAP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003264.2 2 159
          Gene Symbol: SDHAP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003265.3 2 196
          GenBank mRNA AK057553.1 - 2 170
          AK074764.1 - 2 142
          BC053551.1 - 2 187
          BC062438.1 - 2 174
          DA457940.1 - 2 197

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          dehydrogenase oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          tricarboxylic acid cycle
          succinate metabolic process
          nervous system development
          respiratory electron transport chain
          oxidation-reduction process
          succinate dehydrogenase activity
          protein binding
          succinate dehydrogenase (ubiquinone) activity
          flavin adenine dinucleotide binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0