Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGEF9 SIRNA Assays ›
Gene Symbol:
ARHGEF9
Gene Name:
Cdc42 guanine nucleotide exchange factor 9
Gene Aliases:
COLLYBISTIN, EIEE8, HPEM-2, PEM-2, PEM2
Chromosome Location:
Chr.X: 63634967 - 63786025 on Build GRCh38
UniGene ID:
Hs.54697
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22826
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGEF9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001173479.1
NP_001166950.1
3
688
NM_001173480.1
NP_001166951.1
3
1094
NM_015185.2
NP_056000.1
4
1285
XM_005262249.2
XP_005262306.1
4
1347
XM_005262250.3
XP_005262307.1
4
1245
XM_005262251.1
XP_005262308.1
4
595
XM_005262252.1
XP_005262309.1
5
614
XM_011530890.1
XP_011529192.1
5
1029
XM_011530892.1
XP_011529194.1
4
853
XM_017029363.1
XP_016884852.1
5
1374
XM_017029364.1
XP_016884853.1
6
1151
XM_017029365.1
XP_016884854.1
4
1252
XM_017029366.1
XP_016884855.1
5
1033
XM_017029367.1
XP_016884856.1
5
1022
XM_017029368.1
XP_016884857.1
5
1323
XM_017029369.1
XP_016884858.1
4
1347
XM_017029370.1
XP_016884859.1
4
1256
XM_017029371.1
XP_016884860.1
4
1245
XM_017029372.1
XP_016884861.1
4
595
XM_017029373.1
XP_016884862.1
5
1388
XM_017029374.1
XP_016884863.1
3
1076
XM_017029375.1
XP_016884864.1
4
1253
XM_017029376.1
XP_016884865.1
4
1253
XM_017029377.1
XP_016884866.1
4
1256
XM_017029378.1
XP_016884867.1
4
1347
XM_017029379.1
XP_016884868.1
4
595
XM_017029380.1
XP_016884869.1
3
1076
GenBank mRNA
AB007884.1
-
4
1247
AK289993.1
-
4
699
AK295033.1
-
3
700
AK295178.1
-
4
595
AL832116.1
-
4
856
BC117406.1
-
4
596
40125957
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
ion transmembrane transport
regulation of Rho protein signal transduction
positive regulation of apoptotic process
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0