Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ERGIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015966.2 | NP_057050.1 | 6 | 653 | ||
NM_198398.1 | NP_938408.1 | 6 | 653 | |||
GenBank mRNA | AF077030.1 | - | 6 | 611 | ||
AF091085.1 | - | 4 | 264 | |||
AF116614.1 | - | 6 | 760 | |||
AF151812.1 | - | 6 | 611 | |||
AF308298.1 | - | 6 | 613 | |||
AK074550.1 | - | 6 | 601 | |||
AK300916.1 | - | 6 | 602 | |||
AL161963.1 | - | 3 | 294 | |||
BC009765.1 | - | 6 | 607 | 4283844 | ||
BC014014.2 | - | 6 | 606 | 3954269 | ||
BU528333.1 | - | 6 | 630 | |||
CA489637.1 | - | 6 | 653 | |||
EF206690.1 | - | 6 | 611 |