Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291571.1 | NP_001278500.1 | 10 | 1375 | ||
NM_001291572.1 | NP_001278501.1 | 9 | 1312 | |||
NM_005899.4 | NP_005890.2 | 10 | 1375 | |||
NM_031862.3 | NP_114068.1 | 10 | 1183 | |||
XM_011524813.1 | XP_011523115.1 | 10 | 1484 | |||
XM_017024642.1 | XP_016880131.1 | 11 | 2487 | |||
GenBank mRNA | AK128512.1 | - | 2 | 1032 | ||
AK290005.1 | - | 10 | 1131 | |||
AK299328.1 | - | 9 | 1080 | |||
AY450308.1 | - | 10 | 1097 | |||
BC009808.2 | - | 10 | 1102 | 2989212 | ||
D30756.1 | - | 10 | 1173 | |||
X76952.1 | - | 10 | 1125 |