Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144711.5 | NP_653312.2 | 4 | 1790 | ||
GenBank mRNA | AK095131.1 | - | 1366 | |||
BC010437.1 | - | 1624 | 3354344 | |||
BC016950.1 | - | 1 | 82 |
Gene Symbol: PHOSPHO2-KLHL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199290.1 | NP_001186219.1 | 6 | 1921 | ||
GenBank mRNA | AK090653.1 | - | 6 | 1884 | ||
AX746559.1 | - | 6 | 1884 |