Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009619.1 | NP_001009619.1 | 7 | 525 | ||
GenBank mRNA | BC088221.1 | - | 7 | 525 | 7321552 | |
FQ214074.1 | - | 6 | 469 | |||
FQ218444.1 | - | 7 | 538 | |||
FQ220638.1 | - | 7 | 538 | |||
FQ222868.1 | - | 7 | 542 | |||
FQ229330.1 | - | 7 | 541 |