Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195517.1 | NP_001182446.1 | 1, 2 | 801 | ||
NM_005521.3 | NP_005512.1 | 1, 2 | 801 | |||
GenBank mRNA | BC130530.1 | - | 1, 2 | 801 | 40146561 | |
M62626.1 | - | 1, 2 | 1581 | |||
M75952.1 | - | 1, 2 | 575 | |||
S38742.1 | - | 1, 2 | 661 |