Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TNIP3 SIRNA Assays ›
          Gene Symbol:
          TNIP3
          Gene Name:
          TNFAIP3 interacting protein 3
          Gene Aliases:
          ABIN-3, LIND
          Chromosome Location:
          Chr.4: 121127161 - 121227609 on Build GRCh38
          UniGene ID:
          Hs.208206
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 31928

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TNIP3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001128843.2 NP_001122315.2 6, 7 491
          NM_001244764.1 NP_001231693.1 6, 7 531
          NM_024873.5 NP_079149.3 3, 4 436
          XM_011532256.2 XP_011530558.1 7, 8 764
          XM_011532257.2 XP_011530559.1 7, 8 764
          XM_017008619.1 XP_016864108.1 7, 8 764
          XM_017008620.1 XP_016864109.1 7, 8 627
          XM_017008621.1 XP_016864110.1 6, 7 674
          XM_017008622.1 XP_016864111.1 6, 7 927
          XM_017008623.1 XP_016864112.1 6, 7 537
          XM_017008624.1 XP_016864113.1 5, 6 482
          XM_017008625.1 XP_016864114.1 4, 5 1015
          GenBank mRNA AF277289.1 - 3, 4 425
          AJ320534.1 - 3, 4 425
          AK024815.1 - 1, 2 38
          AK290409.1 - 3, 4 417
          AK301058.1 - 6, 7 491
          AK303192.1 - 6, 7 533
          BC128408.1 - 3, 4 424 40115867
          BP335455.1 - 3, 4 436
          DC389224.1 - 6, 7 491
          EF585496.1 - 3, 4 425

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          MyD88-independent toll-like receptor signaling pathway
          inflammatory response
          toll-like receptor 4 signaling pathway
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          cellular response to lipopolysaccharide
          protein binding
          polyubiquitin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-49ts5:80/100.66.72.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0