Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DUSP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304794.1 | NP_001291723.1 | 3 | 1221 | ||
NM_001304795.1 | NP_001291724.1 | 2 | 1054 | |||
NM_001304796.1 | NP_001291725.1 | 3 | 944 | |||
NM_152511.4 | NP_689724.3 | 2 | 1054 | |||
XM_005261368.4 | XP_005261425.1 | 2 | 1054 | |||
XM_006724148.3 | XP_006724211.1 | 3 | 1183 | |||
XM_011529920.2 | XP_011528222.1 | 4 | 1350 | |||
XM_011529921.2 | XP_011528223.1 | 4 | 1290 | |||
XM_017028627.1 | XP_016884116.1 | 3 | 1183 | |||
XM_017028628.1 | XP_016884117.1 | 3 | 1183 | |||
GenBank mRNA | AF461689.1 | - | 2 | 1049 | ||
AF533017.1 | - | 2 | 769 | |||
AK025036.1 | - | 1 | 227 | |||
AK056074.1 | - | 2 | 1049 | |||
BC028724.1 | - | 3 | 908 | |||
BC030987.1 | - | 2 | 1007 | 4730282 | ||
CR456406.1 | - | 1 | 619 |