Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TNF SIRNA Assays ›
          Gene Symbol:
          TNF
          Gene Name:
          tumor necrosis factor
          Gene Aliases:
          DIF, TNF-alpha, TNFA, TNFSF2, TNLG1F
          Chromosome Location:
          Chr.6: 31575567 - 31578336 on Build GRCh38
          UniGene ID:
          Hs.241570
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 4021

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TNF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000594.3 NP_000585.2 2, 3 408
          GenBank mRNA AK314960.1 - 2, 3 407
          BC028148.1 - 2, 3 390 5217636
          BI908079.1 - 2, 3 403
          BP215875.1 - 2, 3 408
          FJ795028.1 - 2, 3 232
          HQ201306.2 - 2, 3 233
          M10988.1 - 2, 3 318
          X01394.1 - 2, 3 385

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus, translocation
          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          activation of MAPKKK activity
          activation of MAPK activity
          positive regulation of cytokine production
          positive regulation of protein phosphorylation
          chronic inflammatory response to antigenic stimulus
          negative regulation of cytokine secretion involved in immune response
          positive regulation of chronic inflammatory response to antigenic stimulus
          positive regulation of humoral immune response mediated by circulating immunoglobulin
          glucose metabolic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          inflammatory response
          humoral immune response
          I-kappaB kinase/NF-kappaB signaling
          JNK cascade
          extrinsic apoptotic signaling pathway via death domain receptors
          intrinsic apoptotic signaling pathway in response to DNA damage
          response to virus
          response to salt stress
          positive regulation of gene expression
          negative regulation of gene expression
          negative regulation of alkaline phosphatase activity
          regulation of tumor necrosis factor-mediated signaling pathway
          negative regulation of lipid storage
          extracellular matrix organization
          osteoclast differentiation
          sequestering of triglyceride
          cortical actin cytoskeleton organization
          positive regulation of protein complex assembly
          positive regulation of fever generation
          lipopolysaccharide-mediated signaling pathway
          negative regulation of interleukin-6 production
          positive regulation of chemokine production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-6 production
          positive regulation of interleukin-8 production
          receptor biosynthetic process
          positive regulation of peptidyl-serine phosphorylation
          tumor necrosis factor-mediated signaling pathway
          positive regulation of heterotypic cell-cell adhesion
          negative regulation of myosin-light-chain-phosphatase activity
          positive regulation of NF-kappaB import into nucleus
          positive regulation of apoptotic process
          positive regulation of programmed cell death
          regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of protein complex disassembly
          positive regulation of protein complex disassembly
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of MAP kinase activity
          protein kinase B signaling
          positive regulation of JUN kinase activity
          negative regulation of growth of symbiont in host
          negative regulation of viral genome replication
          positive regulation of chemokine biosynthetic process
          positive regulation of interleukin-8 biosynthetic process
          positive regulation of nitric oxide biosynthetic process
          negative regulation of fat cell differentiation
          negative regulation of myoblast differentiation
          negative regulation of osteoblast differentiation
          positive regulation of osteoclast differentiation
          positive regulation of cell adhesion
          positive regulation of protein kinase activity
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of translational initiation by iron
          negative regulation of glucose import
          embryonic digestive tract development
          positive regulation of smooth muscle cell proliferation
          positive regulation of cytokine secretion
          positive regulation of phagocytosis
          regulation of insulin secretion
          defense response to Gram-positive bacterium
          leukocyte tethering or rolling
          negative regulation of lipid catabolic process
          regulation of immunoglobulin secretion
          positive regulation of membrane protein ectodomain proteolysis
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of protein transport
          response to glucocorticoid
          positive regulation of NFAT protein import into nucleus
          positive regulation of hair follicle development
          positive regulation of protein kinase B signaling
          positive regulation of vitamin D biosynthetic process
          positive regulation of calcidiol 1-monooxygenase activity
          epithelial cell proliferation involved in salivary gland morphogenesis
          regulation of branching involved in salivary gland morphogenesis
          negative regulation of branching involved in lung morphogenesis
          positive regulation of ERK1 and ERK2 cascade
          cellular response to amino acid stimulus
          cellular response to nicotine
          cellular response to organic cyclic compound
          death-inducing signaling complex assembly
          positive regulation of mononuclear cell migration
          positive regulation of podosome assembly
          establishment of protein localization to plasma membrane
          extrinsic apoptotic signaling pathway
          necroptotic signaling pathway
          positive regulation of NIK/NF-kappaB signaling
          positive regulation of superoxide dismutase activity
          regulation of establishment of endothelial barrier
          negative regulation of bicellular tight junction assembly
          positive regulation of leukocyte adhesion to vascular endothelial cell
          positive regulation of leukocyte adhesion to arterial endothelial cell
          positive regulation of protein localization to cell surface
          positive regulation of ceramide biosynthetic process
          positive regulation of blood microparticle formation
          positive regulation of chemokine (C-X-C motif) ligand 2 production
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          protease binding
          cytokine activity
          tumor necrosis factor receptor binding
          protein binding
          identical protein binding
          transcription regulatory region DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0